Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 120402671-120402930 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK5RAP2
Entrez Gene ID
55755
Gene Name
CDK5 regulatory subunit associated protein 2
Gene Aliases
C48, Cep215, MCPH3
UniGene
Hs.269560
Gene Chromosome Location
Chr.9: 120388869-120580170 on Build GRCh38
Transcripts
NM_001011649.2 NM_001272039.1 NM_018249.5 NR_073554.1 NR_073555.1 NR_073556.1 NR_073557.1 NR_073558.1 XM_006717182.1 XM_006717185.1 XM_011518860.1 XM_017014921.1 XM_017014922.1 XM_017014923.1 XM_017014924.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3847505 c.5248G>A p.E1750K Chr.9: 120402865-120402865 on Build GRCh38
4847422 c.5220G>A p.Q1740Q Chr.9: 120402893-120402893 on Build GRCh38
5965518 c.5183T>C p.L1728P Chr.9: 120402930-120402930 on Build GRCh38
6625552 c.5260C>G p.Q1754E Chr.9: 120402853-120402853 on Build GRCh38
6182485 c.5278A>T p.S1760C Chr.9: 120402835-120402835 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs940306476 Chr.9: 120402820-120402820 on Build GRCh38
rs372996769 Chr.9: 120402784-120402784 on Build GRCh38
rs772132380 Chr.9: 120402803-120402803 on Build GRCh38
rs142554102 Chr.9: 120402890-120402890 on Build GRCh38
rs202060038 Chr.9: 120402748-120402748 on Build GRCh38
rs987083321 Chr.9: 120402796-120402796 on Build GRCh38
rs772661078 Chr.9: 120402788-120402788 on Build GRCh38
rs768628011 Chr.9: 120402815-120402815 on Build GRCh38
rs921392894 Chr.9: 120402749-120402749 on Build GRCh38
rs752959590 Chr.9: 120402756-120402756 on Build GRCh38
rs377272432 Chr.9: 120402830-120402830 on Build GRCh38
rs201889110 Chr.9: 120402883-120402883 on Build GRCh38
rs573792229 Chr.9: 120402729-120402729 on Build GRCh38
rs746748616 Chr.9: 120402811-120402811 on Build GRCh38
rs749431479 Chr.9: 120402782-120402782 on Build GRCh38
rs778505181 Chr.9: 120402728-120402728 on Build GRCh38
rs762693479 Chr.9: 120402847-120402847 on Build GRCh38
rs758686512 Chr.9: 120402902-120402902 on Build GRCh38
rs778396644 Chr.9: 120402776-120402776 on Build GRCh38
rs200081417 Chr.9: 120402751-120402751 on Build GRCh38
rs753148214 Chr.9: 120402872-120402872 on Build GRCh38
rs886063389 Chr.9: 120402878-120402878 on Build GRCh38
rs754398195 Chr.9: 120402891-120402891 on Build GRCh38
rs555457590 Chr.9: 120402719-120402719 on Build GRCh38
rs757776855 Chr.9: 120402898-120402898 on Build GRCh38
rs34561359 Chr.9: 120402739-120402740 on Build GRCh38
rs764731630 Chr.9: 120402826-120402826 on Build GRCh38
rs545646890 Chr.9: 120402856-120402856 on Build GRCh38
rs201584349 Chr.9: 120402785-120402785 on Build GRCh38
rs780115166 Chr.9: 120402905-120402905 on Build GRCh38
rs369647291 Chr.9: 120402906-120402906 on Build GRCh38
rs199918999 Chr.9: 120402818-120402818 on Build GRCh38
rs746111764 Chr.9: 120402786-120402786 on Build GRCh38
rs587783392 Chr.9: 120402886-120402886 on Build GRCh38
rs200521701 Chr.9: 120402757-120402757 on Build GRCh38
rs912780527 Chr.9: 120402801-120402801 on Build GRCh38
rs965481103 Chr.9: 120402734-120402734 on Build GRCh38
rs111436881 Chr.9: 120402834-120402834 on Build GRCh38
rs572551421 Chr.9: 120402901-120402901 on Build GRCh38
rs775386363 Chr.9: 120402810-120402810 on Build GRCh38
rs776538171 Chr.9: 120402817-120402817 on Build GRCh38

View Full Product Details