Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 81151895-81152168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSS3
Entrez Gene ID
79611
Gene Name
acyl-CoA synthetase short-chain family member 3
Gene Aliases
-
UniGene
Hs.259559
Gene Chromosome Location
Chr.12: 81078018-81257795 on Build GRCh38
Transcripts
NM_024560.2 XM_005269150.2 XM_005269151.4 XM_011538740.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
343462 c.1028G>C p.G343A Chr.12: 81152026-81152026 on Build GRCh38
1993470 c.1049A>G p.Y350C Chr.12: 81152047-81152047 on Build GRCh38
1993469 c.1046C>T p.S349F Chr.12: 81152044-81152044 on Build GRCh38
695667 c.1016C>T p.A339V Chr.12: 81152014-81152014 on Build GRCh38
381901 c.1097A>T p.E366V Chr.12: 81152095-81152095 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781414153 Chr.12: 81152075-81152075 on Build GRCh38
rs140829003 Chr.12: 81152031-81152031 on Build GRCh38
rs777300115 Chr.12: 81151981-81151981 on Build GRCh38
rs746611934 Chr.12: 81152104-81152104 on Build GRCh38
rs571077684 Chr.12: 81152108-81152108 on Build GRCh38
rs774950781 Chr.12: 81151975-81151975 on Build GRCh38
rs750005466 Chr.12: 81152067-81152067 on Build GRCh38
rs777212334 Chr.12: 81151926-81151926 on Build GRCh38
rs780328649 Chr.12: 81152085-81152086 on Build GRCh38
rs751711020 Chr.12: 81152026-81152026 on Build GRCh38
rs770393349 Chr.12: 81152129-81152129 on Build GRCh38
rs1055194880 Chr.12: 81151919-81151919 on Build GRCh38
rs778710253 Chr.12: 81152056-81152056 on Build GRCh38
rs753607600 Chr.12: 81152041-81152041 on Build GRCh38
rs769103516 Chr.12: 81151978-81151978 on Build GRCh38
rs773537373 Chr.12: 81152130-81152130 on Build GRCh38
rs764713915 Chr.12: 81151920-81151920 on Build GRCh38
rs762931673 Chr.12: 81152008-81152008 on Build GRCh38
rs867959737 Chr.12: 81152058-81152058 on Build GRCh38
rs779788628 Chr.12: 81152089-81152089 on Build GRCh38
rs773411324 Chr.12: 81152007-81152007 on Build GRCh38
rs757781488 Chr.12: 81152079-81152079 on Build GRCh38
rs548677104 Chr.12: 81151973-81151973 on Build GRCh38
rs762016409 Chr.12: 81151930-81151930 on Build GRCh38
rs757224536 Chr.12: 81152047-81152047 on Build GRCh38
rs755844009 Chr.12: 81151957-81151957 on Build GRCh38
rs199708538 Chr.12: 81151962-81151962 on Build GRCh38
rs368119795 Chr.12: 81151947-81151947 on Build GRCh38
rs766634948 Chr.12: 81152011-81152011 on Build GRCh38
rs772504342 Chr.12: 81152068-81152068 on Build GRCh38
rs771822203 Chr.12: 81151966-81151966 on Build GRCh38
rs756782691 Chr.12: 81151958-81151958 on Build GRCh38
rs752598140 Chr.12: 81151955-81151955 on Build GRCh38
rs747931562 Chr.12: 81151976-81151976 on Build GRCh38
rs747384406 Chr.12: 81151977-81151977 on Build GRCh38
rs781721197 Chr.12: 81152110-81152110 on Build GRCh38
rs748522903 Chr.12: 81152118-81152118 on Build GRCh38
rs746321094 Chr.12: 81151993-81151993 on Build GRCh38
rs972470739 Chr.12: 81152073-81152073 on Build GRCh38
rs3741978 Chr.12: 81151986-81151986 on Build GRCh38
rs766509503 Chr.12: 81151954-81151954 on Build GRCh38
rs563803705 Chr.12: 81151989-81151989 on Build GRCh38
rs200875655 Chr.12: 81152027-81152027 on Build GRCh38
rs201723765 Chr.12: 81151993-81151993 on Build GRCh38
rs750451573 Chr.12: 81151931-81151931 on Build GRCh38
rs559298551 Chr.12: 81151918-81151918 on Build GRCh38
rs779477455 Chr.12: 81151972-81151973 on Build GRCh38

View Full Product Details