Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 80422334-80422607 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM213A
Entrez Gene ID
84293
Gene Name
family with sequence similarity 213 member A
Gene Aliases
C10orf58, PAMM
UniGene
Hs.500333
Gene Chromosome Location
Chr.10: 80407829-80437115 on Build GRCh38
Transcripts
NM_001243778.1 NM_001243779.1 NM_001243780.1 NM_001243781.1 NM_001243782.1 NM_032333.4 XM_011540265.2 XM_011540266.2 XM_017016775.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5613279 c.254G>A p.C85Y Chr.10: 80422492-80422492 on Build GRCh38
4614406 c.212delA p.N73fs*4 Chr.10: 80422450-80422450 on Build GRCh38
6659453 c.237C>T p.A79A Chr.10: 80422475-80422475 on Build GRCh38
4091218 c.221G>C p.G74A Chr.10: 80422459-80422459 on Build GRCh38
5575180 c.242G>A p.R81Q Chr.10: 80422480-80422480 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770698279 Chr.10: 80422506-80422506 on Build GRCh38
rs542292187 Chr.10: 80422453-80422453 on Build GRCh38
rs202047658 Chr.10: 80422541-80422541 on Build GRCh38
rs777110698 Chr.10: 80422385-80422385 on Build GRCh38
rs370909688 Chr.10: 80422475-80422475 on Build GRCh38
rs925551379 Chr.10: 80422586-80422586 on Build GRCh38
rs772924415 Chr.10: 80422503-80422503 on Build GRCh38
rs17105023 Chr.10: 80422583-80422583 on Build GRCh38
rs767536271 Chr.10: 80422531-80422531 on Build GRCh38
rs867682206 Chr.10: 80422474-80422474 on Build GRCh38
rs192099586 Chr.10: 80422545-80422545 on Build GRCh38
rs781529618 Chr.10: 80422487-80422487 on Build GRCh38
rs752526002 Chr.10: 80422536-80422536 on Build GRCh38
rs772693256 Chr.10: 80422392-80422392 on Build GRCh38
rs755839655 Chr.10: 80422450-80422450 on Build GRCh38
rs769020531 Chr.10: 80422381-80422381 on Build GRCh38
rs770653899 Chr.10: 80422391-80422391 on Build GRCh38
rs759353312 Chr.10: 80422413-80422414 on Build GRCh38
rs138058462 Chr.10: 80422479-80422479 on Build GRCh38
rs901686148 Chr.10: 80422585-80422585 on Build GRCh38
rs764927132 Chr.10: 80422571-80422571 on Build GRCh38
rs376415328 Chr.10: 80422409-80422409 on Build GRCh38
rs538801492 Chr.10: 80422393-80422393 on Build GRCh38
rs779214951 Chr.10: 80422459-80422459 on Build GRCh38
rs747448437 Chr.10: 80422378-80422378 on Build GRCh38
rs766527792 Chr.10: 80422588-80422588 on Build GRCh38
rs750569436 Chr.10: 80422462-80422462 on Build GRCh38
rs1870142 Chr.10: 80422540-80422540 on Build GRCh38
rs143142618 Chr.10: 80422523-80422523 on Build GRCh38
rs748568744 Chr.10: 80422495-80422495 on Build GRCh38
rs758475874 Chr.10: 80422471-80422471 on Build GRCh38
rs573121586 Chr.10: 80422391-80422391 on Build GRCh38
rs776416143 Chr.10: 80422399-80422400 on Build GRCh38
rs374415251 Chr.10: 80422377-80422377 on Build GRCh38
rs752573644 Chr.10: 80422439-80422439 on Build GRCh38
rs1870143 Chr.10: 80422572-80422572 on Build GRCh38
rs762639519 Chr.10: 80422504-80422504 on Build GRCh38
rs115389421 Chr.10: 80422552-80422552 on Build GRCh38
rs376644410 Chr.10: 80422560-80422560 on Build GRCh38
rs868836511 Chr.10: 80422561-80422561 on Build GRCh38
rs147085423 Chr.10: 80422476-80422476 on Build GRCh38
rs377622179 Chr.10: 80422456-80422457 on Build GRCh38
rs187412845 Chr.10: 80422400-80422400 on Build GRCh38
rs759609589 Chr.10: 80422412-80422412 on Build GRCh38
rs866201568 Chr.10: 80422500-80422500 on Build GRCh38
rs563809917 Chr.10: 80422516-80422516 on Build GRCh38
rs901689709 Chr.10: 80422482-80422482 on Build GRCh38
rs552990707 Chr.10: 80422370-80422370 on Build GRCh38
rs758677133 Chr.10: 80422550-80422550 on Build GRCh38
rs951944340 Chr.10: 80422513-80422513 on Build GRCh38
rs143497158 Chr.10: 80422499-80422499 on Build GRCh38
rs201451517 Chr.10: 80422430-80422430 on Build GRCh38
rs747087712 Chr.10: 80422584-80422584 on Build GRCh38

View Full Product Details