Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53636783-53637026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPRX
Entrez Gene ID
503834
Gene Name
divergent-paired related homeobox
Gene Aliases
-
UniGene
Hs.579941
Gene Chromosome Location
Chr.19: 53601257-53637057 on Build GRCh38
Transcripts
NM_001012728.1 XM_011527011.2 XM_011527012.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
712870 c.535G>T p.G179C Chr.19: 53636947-53636947 on Build GRCh38
3389325 c.510A>G p.Q170Q Chr.19: 53636922-53636922 on Build GRCh38
3404548 c.373C>T p.R125* Chr.19: 53636785-53636785 on Build GRCh38
712871 c.508C>A p.Q170K Chr.19: 53636920-53636920 on Build GRCh38
319996 c.424A>G p.N142D Chr.19: 53636836-53636836 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866135237 Chr.19: 53636940-53636940 on Build GRCh38
rs780301773 Chr.19: 53636906-53636906 on Build GRCh38
rs184384115 Chr.19: 53636970-53636970 on Build GRCh38
rs372699403 Chr.19: 53636942-53636942 on Build GRCh38
rs937824375 Chr.19: 53636814-53636814 on Build GRCh38
rs761447857 Chr.19: 53636976-53636976 on Build GRCh38
rs10408805 Chr.19: 53636898-53636898 on Build GRCh38
rs370961873 Chr.19: 53636978-53636978 on Build GRCh38
rs370046757 Chr.19: 53636848-53636848 on Build GRCh38
rs773555980 Chr.19: 53636935-53636935 on Build GRCh38
rs769091377 Chr.19: 53636816-53636816 on Build GRCh38
rs747577055 Chr.19: 53636936-53636936 on Build GRCh38
rs773783199 Chr.19: 53636846-53636846 on Build GRCh38
rs367690331 Chr.19: 53636933-53636933 on Build GRCh38
rs763719601 Chr.19: 53636956-53636956 on Build GRCh38
rs565196699 Chr.19: 53636817-53636817 on Build GRCh38
rs1021363380 Chr.19: 53636980-53636980 on Build GRCh38
rs200737613 Chr.19: 53636889-53636889 on Build GRCh38
rs748613654 Chr.19: 53636819-53636819 on Build GRCh38
rs376708716 Chr.19: 53636815-53636815 on Build GRCh38
rs775078581 Chr.19: 53636874-53636874 on Build GRCh38
rs766492524 Chr.19: 53636989-53636989 on Build GRCh38
rs763774653 Chr.19: 53636879-53636879 on Build GRCh38
rs774947764 Chr.19: 53636945-53636945 on Build GRCh38
rs762514061 Chr.19: 53636952-53636952 on Build GRCh38
rs201435914 Chr.19: 53636878-53636878 on Build GRCh38
rs772444739 Chr.19: 53636842-53636842 on Build GRCh38
rs370339375 Chr.19: 53636929-53636929 on Build GRCh38
rs1011027948 Chr.19: 53636972-53636972 on Build GRCh38
rs377492034 Chr.19: 53636977-53636977 on Build GRCh38
rs755247548 Chr.19: 53636928-53636928 on Build GRCh38
rs898877644 Chr.19: 53636834-53636834 on Build GRCh38
rs373577936 Chr.19: 53636899-53636899 on Build GRCh38
rs377588451 Chr.19: 53636904-53636904 on Build GRCh38
rs10409173 Chr.19: 53636914-53636914 on Build GRCh38
rs760665050 Chr.19: 53636987-53636987 on Build GRCh38
rs764762144 Chr.19: 53636853-53636853 on Build GRCh38
rs369622900 Chr.19: 53636893-53636893 on Build GRCh38

View Full Product Details