Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 107949225-107949466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2U1
Entrez Gene ID
113612
Gene Name
cytochrome P450 family 2 subfamily U member 1
Gene Aliases
P450TEC, SPG49, SPG56
UniGene
Hs.109087
Gene Chromosome Location
Chr.4: 107931549-107953461 on Build GRCh38
Transcripts
NM_183075.2 XM_005262717.2 XM_005262720.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7325974 c.1376C>T p.P459L Chr.4: 107949437-107949437 on Build GRCh38
6469205 c.1356C>T p.D452D Chr.4: 107949417-107949417 on Build GRCh38
3598957 c.1396C>T p.R466* Chr.4: 107949457-107949457 on Build GRCh38
3598956 c.1357C>T p.P453S Chr.4: 107949418-107949418 on Build GRCh38
3714792 c.1297G>T p.G433W Chr.4: 107949358-107949358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755199255 Chr.4: 107949361-107949361 on Build GRCh38
rs138472735 Chr.4: 107949438-107949438 on Build GRCh38
rs758463289 Chr.4: 107949352-107949352 on Build GRCh38
rs781535072 Chr.4: 107949380-107949380 on Build GRCh38
rs778388068 Chr.4: 107949393-107949393 on Build GRCh38
rs35849207 Chr.4: 107949291-107949292 on Build GRCh38
rs771661379 Chr.4: 107949411-107949411 on Build GRCh38
rs781763725 Chr.4: 107949282-107949282 on Build GRCh38
rs747965749 Chr.4: 107949437-107949437 on Build GRCh38
rs567414056 Chr.4: 107949360-107949360 on Build GRCh38
rs367948785 Chr.4: 107949331-107949331 on Build GRCh38
rs775157727 Chr.4: 107949416-107949416 on Build GRCh38
rs773404481 Chr.4: 107949426-107949426 on Build GRCh38
rs969943196 Chr.4: 107949326-107949326 on Build GRCh38
rs770176863 Chr.4: 107949404-107949404 on Build GRCh38
rs770180255 Chr.4: 107949384-107949384 on Build GRCh38
rs776264165 Chr.4: 107949434-107949434 on Build GRCh38
rs151162882 Chr.4: 107949266-107949267 on Build GRCh38
rs374076528 Chr.4: 107949328-107949328 on Build GRCh38
rs748563985 Chr.4: 107949292-107949292 on Build GRCh38
rs768478670 Chr.4: 107949422-107949422 on Build GRCh38
rs760397060 Chr.4: 107949419-107949419 on Build GRCh38
rs767302883 Chr.4: 107949382-107949382 on Build GRCh38
rs780128409 Chr.4: 107949354-107949354 on Build GRCh38
rs550643084 Chr.4: 107949320-107949320 on Build GRCh38
rs761638091 Chr.4: 107949303-107949303 on Build GRCh38
rs147100924 Chr.4: 107949394-107949394 on Build GRCh38
rs370357298 Chr.4: 107949367-107949367 on Build GRCh38
rs778626490 Chr.4: 107949285-107949285 on Build GRCh38
rs1041479329 Chr.4: 107949258-107949258 on Build GRCh38
rs748605129 Chr.4: 107949381-107949381 on Build GRCh38
rs945699971 Chr.4: 107949254-107949254 on Build GRCh38

View Full Product Details