Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 40816085-40816356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APBB2
Entrez Gene ID
323
Gene Name
amyloid beta precursor protein binding family B member 2
Gene Aliases
FE65L, FE65L1
UniGene
Hs.479602
Gene Chromosome Location
Chr.4: 40810027-41216714 on Build GRCh38
Transcripts
NM_001166050.1 NM_001166051.1 NM_001166052.1 NM_001166053.1 NM_001166054.1 NM_004307.1 NM_173075.4 XM_005248101.3 XM_006714005.3 XM_006714007.3 XM_006714008.3 XM_006714009.3 XM_006714010.3 XM_006714011.3 XM_006714012.3 XM_011513686.2 XM_011513687.2 XM_017008142.1 XM_017008143.1 XM_017008144.1 XM_017008145.1 XM_017008146.1 XM_017008147.1 XM_017008148.1 XM_017008149.1 XM_017008150.1 XM_017008151.1 XM_017008152.1 XM_017008153.1 XM_017008154.1 XM_017008155.1 XM_017008156.1 XM_017008157.1 XM_017008158.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3603749 c.2114G>A p.R705Q Chr.4: 40816255-40816255 on Build GRCh38
4478368 c.2163C>T p.S721S Chr.4: 40816140-40816140 on Build GRCh38
6246435 c.2240C>A p.T747N Chr.4: 40816129-40816129 on Build GRCh38
2847210 c.2162C>T p.S721F Chr.4: 40816141-40816141 on Build GRCh38
7273242 c.2159G>A p.R720Q Chr.4: 40816210-40816210 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755936257 Chr.4: 40816292-40816292 on Build GRCh38
rs765354981 Chr.4: 40816244-40816244 on Build GRCh38
rs755091636 Chr.4: 40816144-40816144 on Build GRCh38
rs774509520 Chr.4: 40816105-40816105 on Build GRCh38
rs772742835 Chr.4: 40816210-40816210 on Build GRCh38
rs927170379 Chr.4: 40816141-40816141 on Build GRCh38
rs751396112 Chr.4: 40816245-40816245 on Build GRCh38
rs759827967 Chr.4: 40816238-40816238 on Build GRCh38
rs752482685 Chr.4: 40816154-40816154 on Build GRCh38
rs760836576 Chr.4: 40816128-40816128 on Build GRCh38
rs369916931 Chr.4: 40816277-40816277 on Build GRCh38
rs202161633 Chr.4: 40816197-40816197 on Build GRCh38
rs375064552 Chr.4: 40816212-40816212 on Build GRCh38
rs372110290 Chr.4: 40816207-40816207 on Build GRCh38
rs576634338 Chr.4: 40816137-40816137 on Build GRCh38
rs180864438 Chr.4: 40816279-40816279 on Build GRCh38
rs190529086 Chr.4: 40816153-40816153 on Build GRCh38
rs779643865 Chr.4: 40816308-40816308 on Build GRCh38
rs777568610 Chr.4: 40816276-40816276 on Build GRCh38
rs112788816 Chr.4: 40816162-40816162 on Build GRCh38
rs754106736 Chr.4: 40816139-40816139 on Build GRCh38
rs769076200 Chr.4: 40816104-40816104 on Build GRCh38
rs762437843 Chr.4: 40816114-40816114 on Build GRCh38
rs773471496 Chr.4: 40816120-40816120 on Build GRCh38
rs761032631 Chr.4: 40816224-40816224 on Build GRCh38
rs547457464 Chr.4: 40816271-40816271 on Build GRCh38
rs748694088 Chr.4: 40816311-40816311 on Build GRCh38
rs774875932 Chr.4: 40816204-40816204 on Build GRCh38
rs745448879 Chr.4: 40816166-40816166 on Build GRCh38
rs776534558 Chr.4: 40816233-40816233 on Build GRCh38
rs748973343 Chr.4: 40816198-40816198 on Build GRCh38
rs766487790 Chr.4: 40816228-40816228 on Build GRCh38
rs1043939486 Chr.4: 40816171-40816171 on Build GRCh38
rs901349596 Chr.4: 40816319-40816319 on Build GRCh38
rs755636763 Chr.4: 40816190-40816190 on Build GRCh38
rs768150640 Chr.4: 40816201-40816201 on Build GRCh38
rs768084121 Chr.4: 40816115-40816115 on Build GRCh38

View Full Product Details