Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 15002950-15003223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPEB2
Entrez Gene ID
132864
Gene Name
cytoplasmic polyadenylation element binding protein 2
Gene Aliases
CPE-BP2, CPEB-2, hCPEB-2
UniGene
Hs.656937
Gene Chromosome Location
Chr.4: 15002451-15070153 on Build GRCh38
Transcripts
NM_001177381.1 NM_001177382.1 NM_001177383.1 NM_001177384.1 NM_182485.2 NM_182646.2 XM_005248135.3 XM_011513777.2 XM_011513778.2 XM_011513779.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4391412 c.358G>A p.D120N Chr.4: 15003031-15003031 on Build GRCh38
5419965 c.383T>A p.I128N Chr.4: 15003056-15003056 on Build GRCh38
5817392 c.361C>T p.H121Y Chr.4: 15003034-15003034 on Build GRCh38
3732829 c.507_508insGCA p.R169_Q170insA Chr.4: 15003180-15003181 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762669065 Chr.4: 15003180-15003182 on Build GRCh38
rs757980816 Chr.4: 15003057-15003057 on Build GRCh38
rs924129672 Chr.4: 15003166-15003166 on Build GRCh38
rs1005379260 Chr.4: 15003177-15003177 on Build GRCh38
rs754575778 Chr.4: 15003012-15003012 on Build GRCh38
rs1003366790 Chr.4: 15003079-15003079 on Build GRCh38
rs963340728 Chr.4: 15002989-15002989 on Build GRCh38
rs769461026 Chr.4: 15003142-15003142 on Build GRCh38
rs770459742 Chr.4: 15003090-15003090 on Build GRCh38
rs996382261 Chr.4: 15003128-15003128 on Build GRCh38
rs576522935 Chr.4: 15003179-15003180 on Build GRCh38
rs759856423 Chr.4: 15003182-15003183 on Build GRCh38
rs7435318 Chr.4: 15003031-15003031 on Build GRCh38
rs915224335 Chr.4: 15003185-15003185 on Build GRCh38
rs976887423 Chr.4: 15003165-15003165 on Build GRCh38
rs935417543 Chr.4: 15003170-15003170 on Build GRCh38
rs764350501 Chr.4: 15003149-15003149 on Build GRCh38
rs532532287 Chr.4: 15003123-15003123 on Build GRCh38
rs1052155325 Chr.4: 15003104-15003104 on Build GRCh38
rs1014981102 Chr.4: 15003201-15003201 on Build GRCh38
rs935086548 Chr.4: 15003069-15003069 on Build GRCh38
rs779808244 Chr.4: 15003135-15003137 on Build GRCh38
rs747753677 Chr.4: 15003040-15003040 on Build GRCh38
rs765327899 Chr.4: 15003113-15003115 on Build GRCh38
rs745866396 Chr.4: 15003113-15003113 on Build GRCh38
rs756416749 Chr.4: 15003039-15003039 on Build GRCh38
rs758665088 Chr.4: 15003128-15003130 on Build GRCh38
rs767758794 Chr.4: 15003162-15003162 on Build GRCh38
rs750883602 Chr.4: 15003186-15003186 on Build GRCh38
rs947905611 Chr.4: 15003010-15003010 on Build GRCh38
rs768084133 Chr.4: 15003026-15003026 on Build GRCh38
rs965634399 Chr.4: 15003164-15003164 on Build GRCh38
rs1043323498 Chr.4: 15003122-15003122 on Build GRCh38
rs775420996 Chr.4: 15003145-15003145 on Build GRCh38
rs574492698 Chr.4: 15003009-15003009 on Build GRCh38
rs751618952 Chr.4: 15003011-15003011 on Build GRCh38
rs1030935244 Chr.4: 15003130-15003130 on Build GRCh38
rs948041050 Chr.4: 15003187-15003187 on Build GRCh38
rs1014948085 Chr.4: 15002986-15002986 on Build GRCh38
rs1054294649 Chr.4: 15003171-15003171 on Build GRCh38
rs985638365 Chr.4: 15003131-15003131 on Build GRCh38
rs983532354 Chr.4: 15002981-15002981 on Build GRCh38
rs905646796 Chr.4: 15003110-15003110 on Build GRCh38
rs774732907 Chr.4: 15003158-15003158 on Build GRCh38
rs757311727 Chr.4: 15003026-15003027 on Build GRCh38
rs777418031 Chr.4: 15003073-15003073 on Build GRCh38
rs563457090 Chr.4: 15003105-15003105 on Build GRCh38
rs746474044 Chr.4: 15003075-15003075 on Build GRCh38
rs946381888 Chr.4: 15003116-15003116 on Build GRCh38
rs750609126 Chr.4: 15003119-15003127 on Build GRCh38
rs762657552 Chr.4: 15003146-15003146 on Build GRCh38
rs973320401 Chr.4: 15003007-15003007 on Build GRCh38
rs746753686 Chr.4: 15003154-15003155 on Build GRCh38
rs911482032 Chr.4: 15003133-15003133 on Build GRCh38
rs954940112 Chr.4: 15002980-15002980 on Build GRCh38

View Full Product Details