Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36135416-36135657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBR3
Entrez Gene ID
874
Gene Name
carbonyl reductase 3
Gene Aliases
HEL-S-25, SDR21C2, hCBR3
UniGene
Hs.154510
Gene Chromosome Location
Chr.21: 36134901-36146562 on Build GRCh38
Transcripts
NM_001236.3 XM_011529772.2 NR_038892.1 NR_038893.1

Gene Symbol
CBR3-AS1
Entrez Gene ID
100506428
Gene Name
CBR3 antisense RNA 1
Gene Aliases
PlncRNA-1, PlncRNA1
UniGene
Hs.651214
Gene Chromosome Location
Chr.21: 36131767-36156308 on Build GRCh38
Transcripts
NM_001236.3 XM_011529772.2 NR_038892.1 NR_038893.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6620966 c.263T>C p.V88A Chr.21: 36135455-36135455 on Build GRCh38
5086451 c.244delG p.L84fs*26 Chr.21: 36135436-36135436 on Build GRCh38
3379178 c.228C>T p.F76F Chr.21: 36135420-36135420 on Build GRCh38
4134915 c.250C>G p.L84V Chr.21: 36135442-36135442 on Build GRCh38
5619457 c.279C>T p.V93V Chr.21: 36135471-36135471 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs909990926 Chr.21: 36135474-36135474 on Build GRCh38
rs1001246453 Chr.21: 36135544-36135544 on Build GRCh38
rs745795206 Chr.21: 36135435-36135436 on Build GRCh38
rs769675099 Chr.21: 36135490-36135490 on Build GRCh38
rs2835285 Chr.21: 36135469-36135469 on Build GRCh38
rs881711 Chr.21: 36135447-36135447 on Build GRCh38
rs769910236 Chr.21: 36135506-36135506 on Build GRCh38
rs9282628 Chr.21: 36135442-36135442 on Build GRCh38
rs761213324 Chr.21: 36135438-36135438 on Build GRCh38
rs1013862013 Chr.21: 36135574-36135574 on Build GRCh38
rs765355130 Chr.21: 36135441-36135441 on Build GRCh38
rs747878161 Chr.21: 36135488-36135488 on Build GRCh38
rs959664414 Chr.21: 36135552-36135552 on Build GRCh38
rs997859204 Chr.21: 36135623-36135623 on Build GRCh38
rs377655909 Chr.21: 36135439-36135439 on Build GRCh38
rs777245360 Chr.21: 36135533-36135533 on Build GRCh38
rs770729006 Chr.21: 36135507-36135507 on Build GRCh38
rs951649957 Chr.21: 36135636-36135636 on Build GRCh38
rs891701900 Chr.21: 36135554-36135554 on Build GRCh38
rs1010052083 Chr.21: 36135557-36135557 on Build GRCh38
rs754150327 Chr.21: 36135440-36135440 on Build GRCh38
rs881712 Chr.21: 36135471-36135471 on Build GRCh38
rs748907392 Chr.21: 36135505-36135505 on Build GRCh38
rs865839177 Chr.21: 36135615-36135615 on Build GRCh38
rs1042854016 Chr.21: 36135559-36135559 on Build GRCh38
rs1045144596 Chr.21: 36135508-36135508 on Build GRCh38
rs369480097 Chr.21: 36135498-36135498 on Build GRCh38
rs774027205 Chr.21: 36135436-36135436 on Build GRCh38
rs757680105 Chr.21: 36135610-36135610 on Build GRCh38
rs769031361 Chr.21: 36135527-36135527 on Build GRCh38
rs1021646093 Chr.21: 36135577-36135577 on Build GRCh38
rs1048219171 Chr.21: 36135516-36135516 on Build GRCh38
rs1035364485 Chr.21: 36135549-36135549 on Build GRCh38
rs754840426 Chr.21: 36135453-36135453 on Build GRCh38
rs773933523 Chr.21: 36135509-36135509 on Build GRCh38
rs939326024 Chr.21: 36135531-36135531 on Build GRCh38
rs758654657 Chr.21: 36135444-36135444 on Build GRCh38
rs568157218 Chr.21: 36135578-36135578 on Build GRCh38
rs966972159 Chr.21: 36135603-36135603 on Build GRCh38
rs45581835 Chr.21: 36135608-36135608 on Build GRCh38
rs375968532 Chr.21: 36135627-36135627 on Build GRCh38
rs559253237 Chr.21: 36135592-36135592 on Build GRCh38

View Full Product Details