Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 167869632-167869877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GALT1
Entrez Gene ID
8708
Gene Name
beta-1,3-galactosyltransferase 1
Gene Aliases
beta3Gal-T1
UniGene
Hs.735833
Gene Chromosome Location
Chr.2: 167293060-167870856 on Build GRCh38
Transcripts
NM_020981.3 XM_005246931.3 XM_006712819.3 XM_011512085.2 NR_131227.1

Gene Symbol
LOC105616981
Entrez Gene ID
105616981
Gene Name
uncharacterized LOC105616981
Gene Aliases
-
UniGene
Hs.512811
Gene Chromosome Location
Chr.2: 167814774-167941144 on Build GRCh38
Transcripts
NM_020981.3 XM_005246931.3 XM_006712819.3 XM_011512085.2 NR_131227.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3569562 c.628G>T p.G210* Chr.2: 167869667-167869667 on Build GRCh38
1009035 c.716C>T p.T239I Chr.2: 167869755-167869755 on Build GRCh38
5917534 c.731C>T p.S244L Chr.2: 167869770-167869770 on Build GRCh38
4518850 c.835_836CC>TT p.P279F Chr.2: 167869874-167869875 on Build GRCh38
1009034 c.705C>A p.F235L Chr.2: 167869744-167869744 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748943097 Chr.2: 167869663-167869663 on Build GRCh38
rs763607267 Chr.2: 167869790-167869790 on Build GRCh38
rs753270689 Chr.2: 167869777-167869777 on Build GRCh38
rs746868901 Chr.2: 167869688-167869688 on Build GRCh38
rs927223237 Chr.2: 167869703-167869703 on Build GRCh38
rs767958450 Chr.2: 167869775-167869775 on Build GRCh38
rs758231595 Chr.2: 167869813-167869813 on Build GRCh38
rs370442606 Chr.2: 167869774-167869774 on Build GRCh38
rs1041753752 Chr.2: 167869727-167869727 on Build GRCh38
rs774924741 Chr.2: 167869766-167869766 on Build GRCh38
rs745694418 Chr.2: 167869812-167869812 on Build GRCh38
rs143825929 Chr.2: 167869807-167869807 on Build GRCh38
rs148250645 Chr.2: 167869689-167869689 on Build GRCh38
rs114283668 Chr.2: 167869817-167869817 on Build GRCh38
rs150778768 Chr.2: 167869834-167869834 on Build GRCh38
rs773363253 Chr.2: 167869750-167869750 on Build GRCh38
rs771321632 Chr.2: 167869837-167869837 on Build GRCh38
rs753331247 Chr.2: 167869799-167869799 on Build GRCh38
rs141741243 Chr.2: 167869828-167869828 on Build GRCh38
rs893415489 Chr.2: 167869680-167869680 on Build GRCh38
rs144099662 Chr.2: 167869679-167869679 on Build GRCh38
rs756510877 Chr.2: 167869780-167869780 on Build GRCh38
rs561399093 Chr.2: 167869762-167869762 on Build GRCh38
rs746974862 Chr.2: 167869818-167869818 on Build GRCh38
rs759629237 Chr.2: 167869856-167869856 on Build GRCh38
rs778736367 Chr.2: 167869673-167869673 on Build GRCh38
rs774693719 Chr.2: 167869846-167869846 on Build GRCh38
rs749376311 Chr.2: 167869835-167869835 on Build GRCh38
rs35398738 Chr.2: 167869666-167869666 on Build GRCh38
rs569683849 Chr.2: 167869675-167869675 on Build GRCh38
rs778503573 Chr.2: 167869810-167869810 on Build GRCh38

View Full Product Details