Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 17988093-17988364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MGME1
Entrez Gene ID
92667
Gene Name
mitochondrial genome maintenance exonuclease 1
Gene Aliases
C20orf72, DDK1, MTDPS11, bA504H3.4
UniGene
Hs.320823
Gene Chromosome Location
Chr.20: 17968594-17991122 on Build GRCh38
Transcripts
NM_001310338.1 NM_001310339.1 NM_052865.3 XM_005260867.2 XM_005260870.3 XM_006723663.3 XM_011529394.1 XM_011529395.1 XM_017028127.1 XM_017028128.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4806316 c.752A>G p.D251G Chr.20: 17988186-17988186 on Build GRCh38
7182128 c.756G>C p.W252C Chr.20: 17988190-17988190 on Build GRCh38
7271416 c.750T>C p.I250I Chr.20: 17988184-17988184 on Build GRCh38
254590 c.833C>A p.A278D Chr.20: 17988267-17988267 on Build GRCh38
5612812 c.804C>T p.N268N Chr.20: 17988238-17988238 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs76599088 Chr.20: 17988228-17988228 on Build GRCh38
rs28455091 Chr.20: 17988280-17988280 on Build GRCh38
rs776995556 Chr.20: 17988140-17988140 on Build GRCh38
rs772516937 Chr.20: 17988180-17988183 on Build GRCh38
rs766001991 Chr.20: 17988333-17988333 on Build GRCh38
rs1012346043 Chr.20: 17988287-17988287 on Build GRCh38
rs906607042 Chr.20: 17988341-17988341 on Build GRCh38
rs922093654 Chr.20: 17988162-17988162 on Build GRCh38
rs750110660 Chr.20: 17988222-17988222 on Build GRCh38
rs200083126 Chr.20: 17988167-17988167 on Build GRCh38
rs748032328 Chr.20: 17988249-17988249 on Build GRCh38
rs769949021 Chr.20: 17988267-17988267 on Build GRCh38
rs748807451 Chr.20: 17988129-17988129 on Build GRCh38
rs370021658 Chr.20: 17988284-17988284 on Build GRCh38
rs963950130 Chr.20: 17988144-17988144 on Build GRCh38
rs781471759 Chr.20: 17988213-17988213 on Build GRCh38
rs773795690 Chr.20: 17988255-17988256 on Build GRCh38
rs566461651 Chr.20: 17988152-17988152 on Build GRCh38
rs767987313 Chr.20: 17988285-17988285 on Build GRCh38
rs151283958 Chr.20: 17988169-17988169 on Build GRCh38
rs756464444 Chr.20: 17988308-17988308 on Build GRCh38
rs755842621 Chr.20: 17988137-17988137 on Build GRCh38
rs1046438200 Chr.20: 17988322-17988322 on Build GRCh38
rs778728771 Chr.20: 17988248-17988248 on Build GRCh38
rs202011705 Chr.20: 17988241-17988241 on Build GRCh38
rs757637035 Chr.20: 17988195-17988195 on Build GRCh38
rs11551769 Chr.20: 17988211-17988211 on Build GRCh38
rs143218981 Chr.20: 17988276-17988276 on Build GRCh38
rs763057974 Chr.20: 17988163-17988163 on Build GRCh38
rs772022766 Chr.20: 17988252-17988252 on Build GRCh38
rs774845775 Chr.20: 17988283-17988283 on Build GRCh38
rs765382031 Chr.20: 17988142-17988142 on Build GRCh38
rs773091100 Chr.20: 17988254-17988254 on Build GRCh38
rs373097075 Chr.20: 17988289-17988289 on Build GRCh38
rs774337684 Chr.20: 17988131-17988131 on Build GRCh38
rs753471163 Chr.20: 17988336-17988336 on Build GRCh38
rs973633097 Chr.20: 17988148-17988148 on Build GRCh38
rs759790673 Chr.20: 17988141-17988141 on Build GRCh38
rs1047814372 Chr.20: 17988143-17988143 on Build GRCh38
rs779379369 Chr.20: 17988128-17988128 on Build GRCh38
rs761757000 Chr.20: 17988138-17988138 on Build GRCh38
rs760525956 Chr.20: 17988266-17988266 on Build GRCh38
rs923883693 Chr.20: 17988260-17988260 on Build GRCh38
rs985582311 Chr.20: 17988196-17988196 on Build GRCh38
rs142454195 Chr.20: 17988237-17988237 on Build GRCh38

View Full Product Details