Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 14825398-14825671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDNF
Entrez Gene ID
441549
Gene Name
cerebral dopamine neurotrophic factor
Gene Aliases
ARMETL1
UniGene
Hs.559067
Gene Chromosome Location
Chr.10: 14819245-14838073 on Build GRCh38
Transcripts
NM_001029954.2 XM_011519488.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5836597 c.302G>T p.R101L Chr.10: 14825562-14825562 on Build GRCh38
4421487 c.347T>G p.L116R Chr.10: 14825517-14825517 on Build GRCh38
1347007 c.302G>A p.R101H Chr.10: 14825562-14825562 on Build GRCh38
1347008 c.281A>C p.K94T Chr.10: 14825583-14825583 on Build GRCh38
4544193 c.351G>A p.K117K Chr.10: 14825513-14825513 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768956956 Chr.10: 14825477-14825477 on Build GRCh38
rs905802311 Chr.10: 14825555-14825555 on Build GRCh38
rs536621261 Chr.10: 14825464-14825464 on Build GRCh38
rs372113842 Chr.10: 14825439-14825439 on Build GRCh38
rs145772897 Chr.10: 14825563-14825563 on Build GRCh38
rs868122479 Chr.10: 14825519-14825519 on Build GRCh38
rs780392138 Chr.10: 14825475-14825475 on Build GRCh38
rs79576512 Chr.10: 14825444-14825444 on Build GRCh38
rs556612042 Chr.10: 14825471-14825471 on Build GRCh38
rs61843027 Chr.10: 14825558-14825558 on Build GRCh38
rs772496527 Chr.10: 14825620-14825620 on Build GRCh38
rs547534574 Chr.10: 14825424-14825424 on Build GRCh38
rs771086059 Chr.10: 14825532-14825532 on Build GRCh38
rs574212035 Chr.10: 14825611-14825611 on Build GRCh38
rs538582773 Chr.10: 14825538-14825538 on Build GRCh38
rs760669505 Chr.10: 14825509-14825509 on Build GRCh38
rs147031877 Chr.10: 14825593-14825593 on Build GRCh38
rs1002739585 Chr.10: 14825506-14825506 on Build GRCh38
rs753883042 Chr.10: 14825442-14825442 on Build GRCh38
rs148962227 Chr.10: 14825579-14825579 on Build GRCh38
rs940786613 Chr.10: 14825441-14825441 on Build GRCh38
rs749846803 Chr.10: 14825476-14825476 on Build GRCh38
rs139150625 Chr.10: 14825501-14825501 on Build GRCh38
rs201935618 Chr.10: 14825574-14825574 on Build GRCh38
rs147677694 Chr.10: 14825594-14825594 on Build GRCh38
rs369901027 Chr.10: 14825562-14825562 on Build GRCh38
rs751018493 Chr.10: 14825466-14825466 on Build GRCh38
rs372260336 Chr.10: 14825465-14825465 on Build GRCh38
rs368935206 Chr.10: 14825452-14825452 on Build GRCh38
rs376628394 Chr.10: 14825507-14825507 on Build GRCh38
rs543001334 Chr.10: 14825617-14825617 on Build GRCh38
rs776234682 Chr.10: 14825621-14825621 on Build GRCh38
rs892808658 Chr.10: 14825429-14825429 on Build GRCh38
rs755477168 Chr.10: 14825609-14825609 on Build GRCh38
rs558712505 Chr.10: 14825557-14825557 on Build GRCh38
rs373556122 Chr.10: 14825641-14825641 on Build GRCh38
rs759836144 Chr.10: 14825511-14825511 on Build GRCh38
rs142592274 Chr.10: 14825610-14825610 on Build GRCh38
rs772299284 Chr.10: 14825504-14825504 on Build GRCh38

View Full Product Details