Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41690465-41690724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C22orf46
Entrez Gene ID
79640
Gene Name
chromosome 22 open reading frame 46
Gene Aliases
-
UniGene
Hs.517653
Gene Chromosome Location
Chr.22: 41688939-41698136 on Build GRCh38
Transcripts
NM_001142964.1 XM_011530373.2 XM_017028939.1 XM_017028811.1

Gene Symbol
SNU13
Entrez Gene ID
4809
Gene Name
SNU13 homolog, small nuclear ribonucleoprotein (U4/U6.U5)
Gene Aliases
15.5K, FA-1, FA1, NHP2L1, NHPX, OTK27, SNRNP15-5, SPAG12, SSFA1
UniGene
Hs.182255
Gene Chromosome Location
Chr.22: 41673933-41690492 on Build GRCh38
Transcripts
NM_001142964.1 XM_011530373.2 XM_017028939.1 XM_017028811.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7275131 c.13C>T p.L5L Chr.22: 41690640-41690640 on Build GRCh38
7312188 c.4T>G p.L2V Chr.22: 41690631-41690631 on Build GRCh38
1034673 c.16C>A p.L6M Chr.22: 41690643-41690643 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs534936907 Chr.22: 41690538-41690538 on Build GRCh38
rs1037684845 Chr.22: 41690688-41690688 on Build GRCh38
rs34632368 Chr.22: 41690497-41690497 on Build GRCh38
rs918071732 Chr.22: 41690528-41690528 on Build GRCh38
rs907947524 Chr.22: 41690679-41690679 on Build GRCh38
rs992680708 Chr.22: 41690525-41690525 on Build GRCh38
rs188171323 Chr.22: 41690664-41690664 on Build GRCh38
rs939180115 Chr.22: 41690629-41690631 on Build GRCh38
rs992937206 Chr.22: 41690666-41690666 on Build GRCh38
rs935406090 Chr.22: 41690519-41690519 on Build GRCh38
rs774116654 Chr.22: 41690687-41690687 on Build GRCh38
rs183487896 Chr.22: 41690609-41690609 on Build GRCh38
rs940688074 Chr.22: 41690684-41690684 on Build GRCh38
rs766967082 Chr.22: 41690488-41690488 on Build GRCh38
rs770760030 Chr.22: 41690509-41690509 on Build GRCh38
rs1030703864 Chr.22: 41690543-41690543 on Build GRCh38
rs989091880 Chr.22: 41690589-41690589 on Build GRCh38
rs1045333134 Chr.22: 41690603-41690603 on Build GRCh38
rs142064282 Chr.22: 41690542-41690542 on Build GRCh38
rs981021393 Chr.22: 41690616-41690616 on Build GRCh38
rs371329128 Chr.22: 41690533-41690533 on Build GRCh38
rs763484672 Chr.22: 41690633-41690633 on Build GRCh38
rs956395804 Chr.22: 41690582-41690582 on Build GRCh38
rs552967017 Chr.22: 41690602-41690602 on Build GRCh38
rs1029120623 Chr.22: 41690508-41690508 on Build GRCh38
rs760894733 Chr.22: 41690690-41690690 on Build GRCh38
rs927853528 Chr.22: 41690628-41690628 on Build GRCh38
rs1022181649 Chr.22: 41690605-41690605 on Build GRCh38

View Full Product Details