Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34709219-34709490 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL21
Entrez Gene ID
6366
Gene Name
C-C motif chemokine ligand 21
Gene Aliases
6Ckine, CKb9, ECL, SCYA21, SLC, TCA4
UniGene
Hs.57907
Gene Chromosome Location
Chr.9: 34709005-34710167 on Build GRCh38
Transcripts
NM_002989.3 XM_011518004.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6517061 c.400C>T p.P134S Chr.9: 34709399-34709399 on Build GRCh38
1734322 c.380G>A p.R127Q Chr.9: 34709419-34709419 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777242507 Chr.9: 34709356-34709356 on Build GRCh38
rs762022123 Chr.9: 34709399-34709399 on Build GRCh38
rs576393548 Chr.9: 34709414-34709414 on Build GRCh38
rs986119971 Chr.9: 34709389-34709389 on Build GRCh38
rs144158276 Chr.9: 34709394-34709394 on Build GRCh38
rs779274137 Chr.9: 34709451-34709452 on Build GRCh38
rs776029442 Chr.9: 34709370-34709370 on Build GRCh38
rs748637141 Chr.9: 34709457-34709460 on Build GRCh38
rs754971604 Chr.9: 34709423-34709423 on Build GRCh38
rs558718985 Chr.9: 34709307-34709307 on Build GRCh38
rs148496138 Chr.9: 34709419-34709419 on Build GRCh38
rs149966060 Chr.9: 34709393-34709393 on Build GRCh38
rs1015219604 Chr.9: 34709337-34709337 on Build GRCh38
rs140038884 Chr.9: 34709282-34709282 on Build GRCh38
rs200838316 Chr.9: 34709291-34709291 on Build GRCh38
rs11574916 Chr.9: 34709242-34709242 on Build GRCh38
rs755312063 Chr.9: 34709362-34709362 on Build GRCh38
rs375090901 Chr.9: 34709347-34709347 on Build GRCh38
rs1044856935 Chr.9: 34709287-34709287 on Build GRCh38
rs752562720 Chr.9: 34709469-34709469 on Build GRCh38
rs948834710 Chr.9: 34709275-34709275 on Build GRCh38
rs746563633 Chr.9: 34709368-34709368 on Build GRCh38
rs371151997 Chr.9: 34709324-34709324 on Build GRCh38
rs112886735 Chr.9: 34709430-34709430 on Build GRCh38
rs200916713 Chr.9: 34709444-34709444 on Build GRCh38
rs919034923 Chr.9: 34709461-34709461 on Build GRCh38
rs1007358262 Chr.9: 34709402-34709402 on Build GRCh38
rs1003452910 Chr.9: 34709330-34709330 on Build GRCh38
rs770321160 Chr.9: 34709369-34709369 on Build GRCh38
rs1027353013 Chr.9: 34709390-34709390 on Build GRCh38
rs767802042 Chr.9: 34709403-34709403 on Build GRCh38
rs749785685 Chr.9: 34709378-34709378 on Build GRCh38
rs370077092 Chr.9: 34709346-34709346 on Build GRCh38
rs147542625 Chr.9: 34709241-34709241 on Build GRCh38
rs374347595 Chr.9: 34709410-34709410 on Build GRCh38
rs540479251 Chr.9: 34709421-34709421 on Build GRCh38

View Full Product Details