Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 59915713-59915974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2J2
Entrez Gene ID
1573
Gene Name
cytochrome P450 family 2 subfamily J member 2
Gene Aliases
CPJ2, CYPIIJ2
UniGene
Hs.152096
Gene Chromosome Location
Chr.1: 59893308-59926798 on Build GRCh38
Transcripts
NM_000775.3 NR_134981.1 NR_134982.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7177780 c.343C>G p.P115A Chr.1: 59915968-59915968 on Build GRCh38
2237581 c.349C>T p.R117* Chr.1: 59915962-59915962 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs536330616 Chr.1: 59915871-59915871 on Build GRCh38
rs982364633 Chr.1: 59915785-59915785 on Build GRCh38
rs916753993 Chr.1: 59915781-59915781 on Build GRCh38
rs149199403 Chr.1: 59915763-59915763 on Build GRCh38
rs867382179 Chr.1: 59915951-59915951 on Build GRCh38
rs901061984 Chr.1: 59915918-59915918 on Build GRCh38
rs981414428 Chr.1: 59915814-59915814 on Build GRCh38
rs889613999 Chr.1: 59915872-59915872 on Build GRCh38
rs778628591 Chr.1: 59915919-59915919 on Build GRCh38
rs749802524 Chr.1: 59915895-59915895 on Build GRCh38
rs1025612301 Chr.1: 59915946-59915946 on Build GRCh38
rs569299576 Chr.1: 59915939-59915939 on Build GRCh38
rs755076600 Chr.1: 59915749-59915749 on Build GRCh38
rs183855853 Chr.1: 59915782-59915782 on Build GRCh38
rs3738474 Chr.1: 59915912-59915912 on Build GRCh38
rs1056051712 Chr.1: 59915870-59915870 on Build GRCh38
rs947461057 Chr.1: 59915898-59915898 on Build GRCh38
rs952740253 Chr.1: 59915774-59915774 on Build GRCh38
rs547354356 Chr.1: 59915862-59915862 on Build GRCh38
rs11572244 Chr.1: 59915914-59915914 on Build GRCh38
rs928194028 Chr.1: 59915810-59915810 on Build GRCh38
rs11572245 Chr.1: 59915765-59915765 on Build GRCh38
rs200013366 Chr.1: 59915819-59915819 on Build GRCh38
rs762891760 Chr.1: 59915948-59915948 on Build GRCh38
rs935596890 Chr.1: 59915829-59915829 on Build GRCh38
rs200329106 Chr.1: 59915920-59915920 on Build GRCh38
rs1052684017 Chr.1: 59915848-59915848 on Build GRCh38
rs531399488 Chr.1: 59915748-59915748 on Build GRCh38
rs912901377 Chr.1: 59915856-59915856 on Build GRCh38
rs2228113 Chr.1: 59915940-59915940 on Build GRCh38
rs386631617 Chr.1: 59915763-59915765 on Build GRCh38
rs763952565 Chr.1: 59915949-59915949 on Build GRCh38
rs759005313 Chr.1: 59915925-59915925 on Build GRCh38
rs561115820 Chr.1: 59915744-59915744 on Build GRCh38
rs769232293 Chr.1: 59915910-59915910 on Build GRCh38
rs565605402 Chr.1: 59915776-59915776 on Build GRCh38

View Full Product Details