Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 110201489-110201750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC40
Entrez Gene ID
51362
Gene Name
cell division cycle 40
Gene Aliases
EHB3, PRP17, PRPF17
UniGene
Hs.428147
Gene Chromosome Location
Chr.6: 110180421-110232220 on Build GRCh38
Transcripts
NM_015891.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5830215 c.346G>A p.E116K Chr.6: 110201627-110201627 on Build GRCh38
7117304 c.345C>T p.A115A Chr.6: 110201626-110201626 on Build GRCh38
5793955 c.281G>C p.G94A Chr.6: 110201562-110201562 on Build GRCh38
7448602 c.379C>T p.Q127* Chr.6: 110201660-110201660 on Build GRCh38
5404152 c.292C>T p.P98S Chr.6: 110201573-110201573 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368971050 Chr.6: 110201525-110201525 on Build GRCh38
rs34592146 Chr.6: 110201626-110201626 on Build GRCh38
rs144308256 Chr.6: 110201623-110201623 on Build GRCh38
rs199865078 Chr.6: 110201677-110201677 on Build GRCh38
rs780094397 Chr.6: 110201612-110201612 on Build GRCh38
rs992837995 Chr.6: 110201580-110201580 on Build GRCh38
rs769615370 Chr.6: 110201534-110201535 on Build GRCh38
rs1006978693 Chr.6: 110201560-110201560 on Build GRCh38
rs776330115 Chr.6: 110201702-110201702 on Build GRCh38
rs528190920 Chr.6: 110201632-110201632 on Build GRCh38
rs749821792 Chr.6: 110201609-110201609 on Build GRCh38
rs548036861 Chr.6: 110201643-110201643 on Build GRCh38
rs997927971 Chr.6: 110201611-110201611 on Build GRCh38
rs751460541 Chr.6: 110201617-110201617 on Build GRCh38
rs771298480 Chr.6: 110201667-110201667 on Build GRCh38
rs771185478 Chr.6: 110201631-110201631 on Build GRCh38
rs1005958274 Chr.6: 110201523-110201523 on Build GRCh38
rs868457806 Chr.6: 110201641-110201641 on Build GRCh38
rs750271195 Chr.6: 110201591-110201591 on Build GRCh38
rs865853615 Chr.6: 110201625-110201625 on Build GRCh38
rs965098626 Chr.6: 110201593-110201593 on Build GRCh38
rs771017331 Chr.6: 110201521-110201522 on Build GRCh38
rs769236423 Chr.6: 110201699-110201699 on Build GRCh38
rs149591848 Chr.6: 110201570-110201570 on Build GRCh38
rs775858350 Chr.6: 110201685-110201685 on Build GRCh38
rs778044418 Chr.6: 110201588-110201588 on Build GRCh38
rs199575324 Chr.6: 110201534-110201534 on Build GRCh38
rs142985180 Chr.6: 110201534-110201534 on Build GRCh38
rs746303717 Chr.6: 110201651-110201651 on Build GRCh38
rs761118671 Chr.6: 110201688-110201688 on Build GRCh38
rs756992316 Chr.6: 110201537-110201537 on Build GRCh38
rs551121911 Chr.6: 110201522-110201526 on Build GRCh38
rs775621752 Chr.6: 110201535-110201535 on Build GRCh38
rs375400280 Chr.6: 110201627-110201627 on Build GRCh38
rs1017921160 Chr.6: 110201571-110201571 on Build GRCh38
rs925847082 Chr.6: 110201644-110201644 on Build GRCh38
rs201998473 Chr.6: 110201533-110201534 on Build GRCh38

View Full Product Details