Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 181059295-181059544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTNL9
Entrez Gene ID
153579
Gene Name
butyrophilin like 9
Gene Aliases
BTN3, BTN8, VDLS1900
UniGene
Hs.546502
Gene Chromosome Location
Chr.5: 181039875-181061525 on Build GRCh38
Transcripts
NM_152547.4 XM_006714823.3 XM_006714824.3 XM_006714825.3 XM_017009113.1 XM_017009114.1 XM_017009116.1 XM_017009117.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6416511 c.1187G>A p.R396H Chr.5: 181059438-181059438 on Build GRCh38
6579163 c.1112C>T p.T371M Chr.5: 181059366-181059366 on Build GRCh38
6404312 c.1219C>T p.P407S Chr.5: 181059470-181059470 on Build GRCh38
6372776 c.1050C>A p.S350R Chr.5: 181059301-181059301 on Build GRCh38
1620183 c.1286A>T p.E429V Chr.5: 181059540-181059540 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746641215 Chr.5: 181059474-181059474 on Build GRCh38
rs928890805 Chr.5: 181059383-181059383 on Build GRCh38
rs1053422917 Chr.5: 181059421-181059421 on Build GRCh38
rs1002253572 Chr.5: 181059483-181059483 on Build GRCh38
rs879845262 Chr.5: 181059398-181059398 on Build GRCh38
rs553199084 Chr.5: 181059485-181059485 on Build GRCh38
rs759986379 Chr.5: 181059367-181059367 on Build GRCh38
rs768065548 Chr.5: 181059385-181059385 on Build GRCh38
rs768186681 Chr.5: 181059334-181059334 on Build GRCh38
rs201167195 Chr.5: 181059481-181059482 on Build GRCh38
rs557374700 Chr.5: 181059392-181059392 on Build GRCh38
rs943120369 Chr.5: 181059402-181059402 on Build GRCh38
rs902384938 Chr.5: 181059450-181059450 on Build GRCh38
rs893067567 Chr.5: 181059437-181059437 on Build GRCh38
rs1031596223 Chr.5: 181059315-181059315 on Build GRCh38
rs775299854 Chr.5: 181059456-181059456 on Build GRCh38
rs765162228 Chr.5: 181059405-181059405 on Build GRCh38
rs776717470 Chr.5: 181059514-181059514 on Build GRCh38
rs536438792 Chr.5: 181059466-181059466 on Build GRCh38
rs780703157 Chr.5: 181059478-181059478 on Build GRCh38
rs980878173 Chr.5: 181059370-181059370 on Build GRCh38
rs960745086 Chr.5: 181059376-181059376 on Build GRCh38
rs969450640 Chr.5: 181059346-181059346 on Build GRCh38
rs1040157951 Chr.5: 181059403-181059403 on Build GRCh38
rs376707332 Chr.5: 181059506-181059506 on Build GRCh38
rs372187409 Chr.5: 181059409-181059409 on Build GRCh38
rs771499714 Chr.5: 181059487-181059487 on Build GRCh38
rs927944317 Chr.5: 181059372-181059372 on Build GRCh38
rs998536185 Chr.5: 181059408-181059408 on Build GRCh38
rs918721621 Chr.5: 181059331-181059331 on Build GRCh38
rs981683323 Chr.5: 181059380-181059380 on Build GRCh38
rs779675336 Chr.5: 181059326-181059326 on Build GRCh38
rs775772469 Chr.5: 181059386-181059386 on Build GRCh38
rs746350810 Chr.5: 181059315-181059316 on Build GRCh38
rs771585402 Chr.5: 181059351-181059351 on Build GRCh38
rs186444058 Chr.5: 181059404-181059404 on Build GRCh38
rs772542694 Chr.5: 181059505-181059505 on Build GRCh38
rs1011434956 Chr.5: 181059330-181059330 on Build GRCh38
rs747623250 Chr.5: 181059486-181059486 on Build GRCh38
rs776040983 Chr.5: 181059345-181059345 on Build GRCh38
rs750425606 Chr.5: 181059452-181059452 on Build GRCh38
rs761874015 Chr.5: 181059521-181059521 on Build GRCh38
rs200345052 Chr.5: 181059322-181059322 on Build GRCh38
rs972684373 Chr.5: 181059379-181059379 on Build GRCh38
rs764203669 Chr.5: 181059396-181059396 on Build GRCh38
rs746697373 Chr.5: 181059328-181059328 on Build GRCh38
rs749760495 Chr.5: 181059348-181059348 on Build GRCh38
rs760856389 Chr.5: 181059387-181059387 on Build GRCh38
rs753897574 Chr.5: 181059401-181059401 on Build GRCh38
rs951542471 Chr.5: 181059332-181059332 on Build GRCh38
rs1022788426 Chr.5: 181059473-181059473 on Build GRCh38
rs746436109 Chr.5: 181059503-181059503 on Build GRCh38
rs774996266 Chr.5: 181059497-181059497 on Build GRCh38
rs754618093 Chr.5: 181059477-181059477 on Build GRCh38
rs571788589 Chr.5: 181059510-181059510 on Build GRCh38
rs774802017 Chr.5: 181059361-181059361 on Build GRCh38
rs764247268 Chr.5: 181059512-181059512 on Build GRCh38

View Full Product Details