Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 94759716-94759989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf3
Entrez Gene ID
84909
Gene Name
chromosome 9 open reading frame 3
Gene Aliases
AOPEP, AP-O, APO, C90RF3, ONPEP
UniGene
Hs.434253
Gene Chromosome Location
Chr.9: 94726604-95148264 on Build GRCh38
Transcripts
NM_001193329.1 NM_001193331.2 NM_032823.5 XM_005252284.2 XM_006717306.1 XM_011519119.1 XM_011519120.2 XM_011519121.2 XM_011519122.2 XM_011519123.2 XM_011519127.1 XM_011519129.1 XM_011519130.1 XM_011519134.2 XM_017015229.1 XM_017015230.1 XM_017015231.1 XM_017015232.1 XM_017015233.1 XM_017015234.1 XM_017015235.1 XM_017015236.1 XM_017015237.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4139693 c.62T>A p.I21K Chr.9: 94759845-94759845 on Build GRCh38
753828 c.84G>T p.L28L Chr.9: 94759867-94759867 on Build GRCh38
4139692 c.62T>A p.I21K Chr.9: 94759845-94759845 on Build GRCh38
7238518 c.62T>A p.I21K Chr.9: 94759845-94759845 on Build GRCh38
1625267 c.194A>T p.Q65L Chr.9: 94759977-94759977 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766899332 Chr.9: 94759815-94759815 on Build GRCh38
rs781192072 Chr.9: 94759778-94759778 on Build GRCh38
rs765476596 Chr.9: 94759942-94759942 on Build GRCh38
rs772182430 Chr.9: 94759903-94759903 on Build GRCh38
rs749099303 Chr.9: 94759909-94759909 on Build GRCh38
rs759309223 Chr.9: 94759911-94759911 on Build GRCh38
rs774828310 Chr.9: 94759924-94759924 on Build GRCh38
rs750543016 Chr.9: 94759869-94759869 on Build GRCh38
rs745943990 Chr.9: 94759768-94759768 on Build GRCh38
rs758349468 Chr.9: 94759772-94759772 on Build GRCh38
rs769628165 Chr.9: 94759784-94759784 on Build GRCh38
rs772961553 Chr.9: 94759786-94759786 on Build GRCh38
rs758534673 Chr.9: 94759875-94759875 on Build GRCh38
rs774086918 Chr.9: 94759915-94759915 on Build GRCh38
rs776085221 Chr.9: 94759825-94759825 on Build GRCh38
rs373154974 Chr.9: 94759792-94759792 on Build GRCh38
rs545258644 Chr.9: 94759764-94759764 on Build GRCh38
rs762271990 Chr.9: 94759938-94759938 on Build GRCh38
rs774119883 Chr.9: 94759820-94759821 on Build GRCh38
rs748090191 Chr.9: 94759845-94759848 on Build GRCh38
rs760609793 Chr.9: 94759950-94759954 on Build GRCh38
rs201706364 Chr.9: 94759910-94759910 on Build GRCh38
rs765508797 Chr.9: 94759868-94759868 on Build GRCh38
rs779977263 Chr.9: 94759879-94759879 on Build GRCh38
rs759094419 Chr.9: 94759802-94759802 on Build GRCh38
rs764685313 Chr.9: 94759927-94759927 on Build GRCh38
rs141638701 Chr.9: 94759835-94759835 on Build GRCh38
rs773125419 Chr.9: 94759912-94759913 on Build GRCh38
rs761238023 Chr.9: 94759826-94759826 on Build GRCh38
rs200850983 Chr.9: 94759795-94759795 on Build GRCh38
rs761327668 Chr.9: 94759925-94759925 on Build GRCh38
rs944914429 Chr.9: 94759847-94759847 on Build GRCh38
rs777602951 Chr.9: 94759894-94759894 on Build GRCh38
rs553785318 Chr.9: 94759930-94759930 on Build GRCh38
rs375897828 Chr.9: 94759943-94759943 on Build GRCh38
rs756041528 Chr.9: 94759891-94759891 on Build GRCh38
rs148159931 Chr.9: 94759833-94759833 on Build GRCh38
rs766548952 Chr.9: 94759966-94759966 on Build GRCh38
rs749008300 Chr.9: 94759790-94759790 on Build GRCh38
rs771658703 Chr.9: 94759922-94759922 on Build GRCh38
rs745464270 Chr.9: 94759921-94759921 on Build GRCh38
rs747989053 Chr.9: 94759783-94759783 on Build GRCh38
rs927597968 Chr.9: 94759800-94759800 on Build GRCh38

View Full Product Details