Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 85286754-85287027 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALX1
Entrez Gene ID
8092
Gene Name
ALX homeobox 1
Gene Aliases
CART1, FND3, HEL23
UniGene
Hs.41683
Gene Chromosome Location
Chr.12: 85280258-85301784 on Build GRCh38
Transcripts
NM_006982.2 XM_011538782.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1476979 c.648C>A p.D216E Chr.12: 85286969-85286969 on Build GRCh38
145091 c.594A>T p.Q198H Chr.12: 85286915-85286915 on Build GRCh38
7082130 c.621T>C p.Y207Y Chr.12: 85286942-85286942 on Build GRCh38
1364641 c.596C>T p.A199V Chr.12: 85286917-85286917 on Build GRCh38
1994430 c.638C>A p.P213Q Chr.12: 85286959-85286959 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048186835 Chr.12: 85286890-85286890 on Build GRCh38
rs778396653 Chr.12: 85286895-85286895 on Build GRCh38
rs772128403 Chr.12: 85286923-85286923 on Build GRCh38
rs758426051 Chr.12: 85286909-85286909 on Build GRCh38
rs781627744 Chr.12: 85286855-85286855 on Build GRCh38
rs367954668 Chr.12: 85286818-85286818 on Build GRCh38
rs115409839 Chr.12: 85286951-85286951 on Build GRCh38
rs950686570 Chr.12: 85286899-85286899 on Build GRCh38
rs577761512 Chr.12: 85286948-85286948 on Build GRCh38
rs888180516 Chr.12: 85286901-85286901 on Build GRCh38
rs751238742 Chr.12: 85286827-85286827 on Build GRCh38
rs1030258242 Chr.12: 85286896-85286896 on Build GRCh38
rs144593505 Chr.12: 85286917-85286917 on Build GRCh38
rs1057393103 Chr.12: 85286784-85286784 on Build GRCh38
rs757136525 Chr.12: 85286894-85286894 on Build GRCh38
rs754715521 Chr.12: 85286992-85286992 on Build GRCh38
rs998096551 Chr.12: 85286969-85286969 on Build GRCh38
rs765267498 Chr.12: 85286946-85286946 on Build GRCh38
rs150997122 Chr.12: 85286868-85286868 on Build GRCh38
rs771162169 Chr.12: 85286916-85286916 on Build GRCh38
rs112505557 Chr.12: 85286979-85286979 on Build GRCh38
rs79907632 Chr.12: 85286918-85286918 on Build GRCh38
rs776549338 Chr.12: 85286932-85286932 on Build GRCh38
rs189121824 Chr.12: 85286836-85286836 on Build GRCh38
rs751012467 Chr.12: 85286991-85286991 on Build GRCh38
rs140886664 Chr.12: 85286897-85286897 on Build GRCh38
rs765874556 Chr.12: 85286977-85286977 on Build GRCh38
rs915604756 Chr.12: 85286804-85286804 on Build GRCh38
rs761480438 Chr.12: 85286938-85286938 on Build GRCh38
rs900348786 Chr.12: 85286823-85286823 on Build GRCh38

View Full Product Details