Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 62833973-62834212 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG4C
Entrez Gene ID
84938
Gene Name
autophagy related 4C cysteine peptidase
Gene Aliases
APG4-C, APG4C, AUTL1, AUTL3
UniGene
Hs.7353
Gene Chromosome Location
Chr.1: 62784106-62865270 on Build GRCh38
Transcripts
NM_032852.3 NM_178221.2 XM_005271288.2 XM_011542313.2 XM_017002600.1 XM_017002601.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5784601 c.1009C>G p.Q337E Chr.1: 62834113-62834113 on Build GRCh38
5784602 c.241C>G p.Q81E Chr.1: 62834113-62834113 on Build GRCh38
2238574 c.963T>C p.G321G Chr.1: 62834067-62834067 on Build GRCh38
4240348 c.186T>C p.Y62Y Chr.1: 62834058-62834058 on Build GRCh38
2238568 c.954T>C p.Y318Y Chr.1: 62834058-62834058 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763347056 Chr.1: 62834076-62834076 on Build GRCh38
rs754043891 Chr.1: 62834136-62834136 on Build GRCh38
rs767637818 Chr.1: 62834102-62834102 on Build GRCh38
rs770410815 Chr.1: 62834024-62834025 on Build GRCh38
rs758186492 Chr.1: 62834137-62834137 on Build GRCh38
rs770427190 Chr.1: 62834059-62834059 on Build GRCh38
rs762589573 Chr.1: 62834025-62834025 on Build GRCh38
rs750542551 Chr.1: 62834104-62834104 on Build GRCh38
rs775679166 Chr.1: 62834072-62834072 on Build GRCh38
rs551767003 Chr.1: 62834059-62834060 on Build GRCh38
rs921454236 Chr.1: 62834181-62834181 on Build GRCh38
rs776125932 Chr.1: 62834062-62834062 on Build GRCh38
rs752098724 Chr.1: 62834088-62834088 on Build GRCh38
rs542777807 Chr.1: 62834024-62834024 on Build GRCh38
rs746886603 Chr.1: 62834147-62834147 on Build GRCh38
rs777668660 Chr.1: 62834012-62834012 on Build GRCh38
rs759056180 Chr.1: 62834068-62834068 on Build GRCh38
rs769511318 Chr.1: 62834069-62834069 on Build GRCh38
rs771858498 Chr.1: 62834003-62834003 on Build GRCh38
rs543316907 Chr.1: 62834167-62834167 on Build GRCh38
rs759118785 Chr.1: 62834064-62834064 on Build GRCh38
rs762476436 Chr.1: 62834097-62834097 on Build GRCh38
rs146422144 Chr.1: 62834080-62834080 on Build GRCh38
rs767334929 Chr.1: 62834130-62834131 on Build GRCh38
rs1033035891 Chr.1: 62834079-62834079 on Build GRCh38
rs565191939 Chr.1: 62834186-62834186 on Build GRCh38
rs1023830177 Chr.1: 62834099-62834099 on Build GRCh38
rs183634447 Chr.1: 62834116-62834116 on Build GRCh38
rs777615519 Chr.1: 62834140-62834140 on Build GRCh38
rs990922751 Chr.1: 62834058-62834059 on Build GRCh38
rs1049187683 Chr.1: 62834075-62834075 on Build GRCh38

View Full Product Details