Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 73407893-73408150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX43
Entrez Gene ID
55510
Gene Name
DEAD-box helicase 43
Gene Aliases
CT13, HAGE
UniGene
Hs.125507
Gene Chromosome Location
Chr.6: 73394562-73417566 on Build GRCh38
Transcripts
NM_018665.2 XM_011535926.1 XM_011535927.2 XM_011535928.2 XM_017011003.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6107768 c.1164G>A p.K388K Chr.6: 73408086-73408086 on Build GRCh38
3928597 c.1144A>G p.S382G Chr.6: 73408066-73408066 on Build GRCh38
1754920 c.1062T>C p.N354N Chr.6: 73407984-73407984 on Build GRCh38
1081537 c.1127T>C p.L376S Chr.6: 73408049-73408049 on Build GRCh38
1445830 c.1087delA p.G365fs*2 Chr.6: 73408009-73408009 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749336993 Chr.6: 73408037-73408037 on Build GRCh38
rs371608743 Chr.6: 73408074-73408074 on Build GRCh38
rs761677993 Chr.6: 73407961-73407961 on Build GRCh38
rs765162298 Chr.6: 73407964-73407964 on Build GRCh38
rs979814638 Chr.6: 73407979-73407979 on Build GRCh38
rs757925484 Chr.6: 73407972-73407972 on Build GRCh38
rs770897683 Chr.6: 73408039-73408039 on Build GRCh38
rs751146932 Chr.6: 73408081-73408081 on Build GRCh38
rs780401899 Chr.6: 73407965-73407965 on Build GRCh38
rs748777346 Chr.6: 73407990-73407990 on Build GRCh38
rs778318746 Chr.6: 73408121-73408121 on Build GRCh38
rs1046678965 Chr.6: 73408031-73408031 on Build GRCh38
rs201225104 Chr.6: 73408075-73408075 on Build GRCh38
rs533108607 Chr.6: 73407983-73407983 on Build GRCh38
rs756575224 Chr.6: 73408118-73408118 on Build GRCh38
rs918036053 Chr.6: 73408099-73408099 on Build GRCh38
rs369032631 Chr.6: 73408067-73408067 on Build GRCh38
rs565631306 Chr.6: 73407945-73407945 on Build GRCh38
rs141846637 Chr.6: 73408041-73408041 on Build GRCh38
rs778459760 Chr.6: 73408030-73408030 on Build GRCh38
rs775450124 Chr.6: 73408113-73408114 on Build GRCh38
rs753059183 Chr.6: 73408115-73408115 on Build GRCh38
rs769992603 Chr.6: 73407928-73407928 on Build GRCh38
rs748539370 Chr.6: 73408025-73408025 on Build GRCh38
rs751141466 Chr.6: 73407994-73407994 on Build GRCh38
rs192224280 Chr.6: 73408042-73408042 on Build GRCh38
rs765792006 Chr.6: 73408079-73408079 on Build GRCh38
rs150607071 Chr.6: 73408048-73408048 on Build GRCh38
rs776354264 Chr.6: 73408057-73408057 on Build GRCh38
rs756626330 Chr.6: 73408027-73408027 on Build GRCh38
rs754727706 Chr.6: 73408016-73408016 on Build GRCh38
rs781398896 Chr.6: 73408024-73408024 on Build GRCh38
rs767209509 Chr.6: 73408084-73408084 on Build GRCh38

View Full Product Details