Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206116892-206116647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AVPR1B
Entrez Gene ID
553
Gene Name
arginine vasopressin receptor 1B
Gene Aliases
AVPR3
UniGene
Hs.1372
Gene Chromosome Location
Chr.1: 206109849-206117048 on Build GRCh38
Transcripts
NM_000707.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6608974 c.85C>T p.R29W Chr.1: 206116806-206116806 on Build GRCh38
6608971 c.75C>A p.P25P Chr.1: 206116816-206116816 on Build GRCh38
3864272 c.84C>T p.G28G Chr.1: 206116807-206116807 on Build GRCh38
3849467 c.107T>C p.V36A Chr.1: 206116784-206116784 on Build GRCh38
6508562 c.242T>A p.L81Q Chr.1: 206116649-206116649 on Build GRCh38
View More

View Full Product Details