Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 31983324-31983551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1551
Entrez Gene ID
55196
Gene Name
KIAA1551
Gene Aliases
C12orf35, UTA2-1
UniGene
Hs.445129
Gene Chromosome Location
Chr.12: 31958800-31993107 on Build GRCh38
Transcripts
NM_018169.3 XM_005253405.2 XM_006719099.2 XM_011520722.1 XM_017019550.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4430934 c.2405A>G p.N802S Chr.12: 31983360-31983360 on Build GRCh38
1235969 c.2461C>T p.P821S Chr.12: 31983416-31983416 on Build GRCh38
2205731 c.2583A>G p.K861K Chr.12: 31983538-31983538 on Build GRCh38
5922952 c.2369C>T p.P790L Chr.12: 31983324-31983324 on Build GRCh38
3811880 c.2380G>T p.E794* Chr.12: 31983335-31983335 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767170229 Chr.12: 31983438-31983438 on Build GRCh38
rs747103032 Chr.12: 31983410-31983410 on Build GRCh38
rs150039507 Chr.12: 31983378-31983378 on Build GRCh38
rs376977882 Chr.12: 31983436-31983436 on Build GRCh38
rs370608026 Chr.12: 31983416-31983416 on Build GRCh38
rs770031561 Chr.12: 31983417-31983417 on Build GRCh38
rs771947480 Chr.12: 31983512-31983512 on Build GRCh38
rs558126712 Chr.12: 31983364-31983364 on Build GRCh38
rs146018580 Chr.12: 31983505-31983505 on Build GRCh38
rs1048632067 Chr.12: 31983453-31983453 on Build GRCh38
rs781173622 Chr.12: 31983493-31983493 on Build GRCh38
rs770866997 Chr.12: 31983411-31983411 on Build GRCh38
rs35398302 Chr.12: 31983520-31983521 on Build GRCh38
rs745944281 Chr.12: 31983497-31983497 on Build GRCh38
rs12297628 Chr.12: 31983386-31983386 on Build GRCh38
rs969693174 Chr.12: 31983494-31983494 on Build GRCh38
rs778272686 Chr.12: 31983471-31983471 on Build GRCh38
rs1041163840 Chr.12: 31983504-31983504 on Build GRCh38
rs761887952 Chr.12: 31983352-31983352 on Build GRCh38
rs1028854599 Chr.12: 31983519-31983519 on Build GRCh38
rs376553451 Chr.12: 31983401-31983401 on Build GRCh38
rs778184597 Chr.12: 31983404-31983404 on Build GRCh38
rs371141683 Chr.12: 31983507-31983507 on Build GRCh38
rs140877209 Chr.12: 31983478-31983478 on Build GRCh38
rs767546025 Chr.12: 31983353-31983353 on Build GRCh38
rs772143497 Chr.12: 31983433-31983433 on Build GRCh38
rs757747299 Chr.12: 31983486-31983486 on Build GRCh38
rs1024932218 Chr.12: 31983485-31983485 on Build GRCh38
rs766072507 Chr.12: 31983370-31983370 on Build GRCh38
rs200548938 Chr.12: 31983423-31983423 on Build GRCh38
rs760259544 Chr.12: 31983435-31983435 on Build GRCh38
rs779185215 Chr.12: 31983391-31983391 on Build GRCh38
rs748491514 Chr.12: 31983398-31983398 on Build GRCh38
rs759160656 Chr.12: 31983441-31983441 on Build GRCh38
rs996868297 Chr.12: 31983515-31983515 on Build GRCh38
rs750023477 Chr.12: 31983356-31983356 on Build GRCh38
rs781507968 Chr.12: 31983414-31983414 on Build GRCh38
rs753128326 Chr.12: 31983467-31983467 on Build GRCh38
rs753358660 Chr.12: 31983440-31983440 on Build GRCh38
rs752325733 Chr.12: 31983491-31983491 on Build GRCh38
rs773228085 Chr.12: 31983434-31983434 on Build GRCh38
rs998874573 Chr.12: 31983418-31983418 on Build GRCh38
rs769973447 Chr.12: 31983501-31983501 on Build GRCh38
rs554538448 Chr.12: 31983419-31983419 on Build GRCh38
rs1004425105 Chr.12: 31983488-31983488 on Build GRCh38
rs758703601 Chr.12: 31983470-31983470 on Build GRCh38
rs764858736 Chr.12: 31983462-31983462 on Build GRCh38

View Full Product Details