Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186544183-186544456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYGS
Entrez Gene ID
1427
Gene Name
crystallin gamma S
Gene Aliases
CRYG8, CTRCT20
UniGene
Hs.376209
Gene Chromosome Location
Chr.3: 186538443-186544378 on Build GRCh38
Transcripts
NM_017541.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5611687 c.12T>A p.T4T Chr.3: 186544315-186544315 on Build GRCh38
4851467 c.14G>C p.G5A Chr.3: 186544313-186544313 on Build GRCh38
479817 c.3G>A p.M1I Chr.3: 186544324-186544324 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs893030750 Chr.3: 186544238-186544238 on Build GRCh38
rs377530357 Chr.3: 186544303-186544303 on Build GRCh38
rs1040551693 Chr.3: 186544412-186544412 on Build GRCh38
rs946753938 Chr.3: 186544381-186544381 on Build GRCh38
rs16860883 Chr.3: 186544274-186544274 on Build GRCh38
rs771176383 Chr.3: 186544275-186544275 on Build GRCh38
rs753106346 Chr.3: 186544369-186544369 on Build GRCh38
rs986069828 Chr.3: 186544396-186544396 on Build GRCh38
rs200400904 Chr.3: 186544344-186544344 on Build GRCh38
rs778117851 Chr.3: 186544373-186544373 on Build GRCh38
rs553020086 Chr.3: 186544330-186544330 on Build GRCh38
rs763911313 Chr.3: 186544336-186544336 on Build GRCh38
rs768134063 Chr.3: 186544244-186544245 on Build GRCh38
rs755427034 Chr.3: 186544261-186544261 on Build GRCh38
rs910658272 Chr.3: 186544400-186544400 on Build GRCh38
rs771855592 Chr.3: 186544309-186544309 on Build GRCh38
rs944893091 Chr.3: 186544214-186544214 on Build GRCh38
rs73055398 Chr.3: 186544252-186544252 on Build GRCh38
rs762968299 Chr.3: 186544338-186544338 on Build GRCh38
rs147700791 Chr.3: 186544343-186544343 on Build GRCh38
rs777407555 Chr.3: 186544279-186544279 on Build GRCh38
rs749752660 Chr.3: 186544374-186544374 on Build GRCh38
rs773393004 Chr.3: 186544337-186544337 on Build GRCh38
rs978188108 Chr.3: 186544415-186544415 on Build GRCh38
rs371031485 Chr.3: 186544372-186544372 on Build GRCh38
rs558458882 Chr.3: 186544259-186544259 on Build GRCh38
rs541307688 Chr.3: 186544289-186544289 on Build GRCh38
rs370988598 Chr.3: 186544316-186544316 on Build GRCh38
rs1034844030 Chr.3: 186544351-186544351 on Build GRCh38
rs759634736 Chr.3: 186544408-186544408 on Build GRCh38
rs756745787 Chr.3: 186544378-186544378 on Build GRCh38
rs563445384 Chr.3: 186544366-186544366 on Build GRCh38
rs956878374 Chr.3: 186544355-186544355 on Build GRCh38
rs370168399 Chr.3: 186544295-186544295 on Build GRCh38
rs770665086 Chr.3: 186544292-186544292 on Build GRCh38
rs370806464 Chr.3: 186544357-186544357 on Build GRCh38
rs112261986 Chr.3: 186544332-186544332 on Build GRCh38

View Full Product Details