Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15427689-15427950 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EAF1
Entrez Gene ID
85403
Gene Name
ELL associated factor 1
Gene Aliases
-
UniGene
Hs.716733
Gene Chromosome Location
Chr.3: 15427557-15442613 on Build GRCh38
Transcripts
NM_033083.6 XM_005265518.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1204852 c.13G>A p.A5T Chr.3: 15427792-15427792 on Build GRCh38
445837 c.69G>A p.E23E Chr.3: 15427848-15427848 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs972272054 Chr.3: 15427897-15427897 on Build GRCh38
rs952413115 Chr.3: 15427917-15427917 on Build GRCh38
rs892477686 Chr.3: 15427789-15427789 on Build GRCh38
rs762752600 Chr.3: 15427877-15427877 on Build GRCh38
rs966429544 Chr.3: 15427848-15427848 on Build GRCh38
rs369264153 Chr.3: 15427916-15427916 on Build GRCh38
rs41284029 Chr.3: 15427876-15427876 on Build GRCh38
rs748304883 Chr.3: 15427820-15427820 on Build GRCh38
rs759842708 Chr.3: 15427749-15427749 on Build GRCh38
rs548787128 Chr.3: 15427729-15427729 on Build GRCh38
rs142692553 Chr.3: 15427741-15427741 on Build GRCh38
rs760602660 Chr.3: 15427902-15427902 on Build GRCh38
rs1000062227 Chr.3: 15427819-15427819 on Build GRCh38
rs764506619 Chr.3: 15427712-15427713 on Build GRCh38
rs376417139 Chr.3: 15427754-15427754 on Build GRCh38
rs747519706 Chr.3: 15427777-15427777 on Build GRCh38
rs764077366 Chr.3: 15427908-15427908 on Build GRCh38
rs569693223 Chr.3: 15427735-15427735 on Build GRCh38
rs1010756524 Chr.3: 15427821-15427821 on Build GRCh38
rs755228974 Chr.3: 15427741-15427741 on Build GRCh38
rs910610505 Chr.3: 15427724-15427724 on Build GRCh38
rs781405243 Chr.3: 15427808-15427808 on Build GRCh38
rs142360478 Chr.3: 15427914-15427914 on Build GRCh38
rs372062063 Chr.3: 15427751-15427751 on Build GRCh38
rs199920203 Chr.3: 15427763-15427763 on Build GRCh38
rs370022222 Chr.3: 15427790-15427790 on Build GRCh38
rs769876525 Chr.3: 15427833-15427833 on Build GRCh38
rs775341002 Chr.3: 15427901-15427901 on Build GRCh38
rs1040334601 Chr.3: 15427744-15427744 on Build GRCh38
rs904339006 Chr.3: 15427816-15427816 on Build GRCh38
rs961727953 Chr.3: 15427892-15427892 on Build GRCh38
rs372055507 Chr.3: 15427715-15427715 on Build GRCh38
rs77636884 Chr.3: 15427860-15427860 on Build GRCh38
rs1041694440 Chr.3: 15427796-15427796 on Build GRCh38
rs539822287 Chr.3: 15427711-15427711 on Build GRCh38
rs780508737 Chr.3: 15427895-15427896 on Build GRCh38
rs575466609 Chr.3: 15427791-15427791 on Build GRCh38
rs554473608 Chr.3: 15427873-15427873 on Build GRCh38

View Full Product Details