Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31143249-31143498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCHCR1
Entrez Gene ID
54535
Gene Name
coiled-coil alpha-helical rod protein 1
Gene Aliases
C6orf18, HCR, SBP
UniGene
Hs.485075
Gene Chromosome Location
Chr.6: 31142439-31158238 on Build GRCh38
Transcripts
NM_001105563.1 NM_001105564.1 NM_019052.3 XM_011514702.2 XM_011514703.1 XM_011514704.2 XM_011514705.1 XM_017010961.1 XM_017010962.1 XM_017010963.1 XM_017010964.1 XM_017010965.1 XM_017010966.1 XM_017010967.1 XM_017010968.1 XM_017010969.1 XM_017010970.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5654063 c.2246A>T p.E749V Chr.6: 31143335-31143335 on Build GRCh38
5654064 c.1979A>T p.E660V Chr.6: 31143335-31143335 on Build GRCh38
4421483 c.1955A>G p.Q652R Chr.6: 31143359-31143359 on Build GRCh38
3684200 c.1966C>T p.R656C Chr.6: 31143348-31143348 on Build GRCh38
3761824 c.2178A>G p.L726L Chr.6: 31143403-31143403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765334394 Chr.6: 31143309-31143326 on Build GRCh38
rs749612470 Chr.6: 31143398-31143398 on Build GRCh38
rs769626815 Chr.6: 31143438-31143438 on Build GRCh38
rs990785024 Chr.6: 31143430-31143430 on Build GRCh38
rs766913794 Chr.6: 31143447-31143447 on Build GRCh38
rs776682186 Chr.6: 31143364-31143364 on Build GRCh38
rs780312235 Chr.6: 31143328-31143328 on Build GRCh38
rs751269045 Chr.6: 31143381-31143381 on Build GRCh38
rs73397100 Chr.6: 31143348-31143348 on Build GRCh38
rs758012592 Chr.6: 31143289-31143289 on Build GRCh38
rs985619853 Chr.6: 31143471-31143471 on Build GRCh38
rs756636243 Chr.6: 31143391-31143391 on Build GRCh38
rs116969494 Chr.6: 31143371-31143371 on Build GRCh38
rs775782643 Chr.6: 31143318-31143332 on Build GRCh38
rs969226922 Chr.6: 31143437-31143437 on Build GRCh38
rs760599372 Chr.6: 31143294-31143294 on Build GRCh38
rs112520959 Chr.6: 31143299-31143299 on Build GRCh38
rs756823571 Chr.6: 31143300-31143300 on Build GRCh38
rs763604842 Chr.6: 31143291-31143291 on Build GRCh38
rs150789792 Chr.6: 31143314-31143314 on Build GRCh38
rs762597318 Chr.6: 31143368-31143368 on Build GRCh38
rs372934272 Chr.6: 31143401-31143401 on Build GRCh38
rs746420594 Chr.6: 31143312-31143312 on Build GRCh38
rs748224886 Chr.6: 31143357-31143357 on Build GRCh38
rs371850031 Chr.6: 31143420-31143420 on Build GRCh38
rs759580927 Chr.6: 31143365-31143365 on Build GRCh38
rs979552601 Chr.6: 31143477-31143477 on Build GRCh38
rs1040589922 Chr.6: 31143329-31143329 on Build GRCh38
rs749477531 Chr.6: 31143330-31143330 on Build GRCh38
rs902697271 Chr.6: 31143301-31143301 on Build GRCh38
rs570015408 Chr.6: 31143302-31143302 on Build GRCh38
rs570821434 Chr.6: 31143366-31143366 on Build GRCh38
rs570019609 Chr.6: 31143347-31143347 on Build GRCh38
rs374842942 Chr.6: 31143429-31143429 on Build GRCh38
rs762878831 Chr.6: 31143347-31143380 on Build GRCh38
rs755429547 Chr.6: 31143400-31143400 on Build GRCh38
rs752357041 Chr.6: 31143282-31143282 on Build GRCh38
rs767399922 Chr.6: 31143358-31143358 on Build GRCh38
rs130073 Chr.6: 31143403-31143403 on Build GRCh38
rs745746346 Chr.6: 31143435-31143435 on Build GRCh38
rs757621536 Chr.6: 31143404-31143404 on Build GRCh38
rs748422751 Chr.6: 31143402-31143402 on Build GRCh38
rs946766056 Chr.6: 31143474-31143474 on Build GRCh38
rs879023466 Chr.6: 31143478-31143478 on Build GRCh38
rs130074 Chr.6: 31143397-31143397 on Build GRCh38
rs765167770 Chr.6: 31143278-31143278 on Build GRCh38
rs760608377 Chr.6: 31143382-31143382 on Build GRCh38
rs905766836 Chr.6: 31143392-31143392 on Build GRCh38

View Full Product Details