Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63763667-63763896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf95
Entrez Gene ID
65998
Gene Name
chromosome 11 open reading frame 95
Gene Aliases
-
UniGene
Hs.191073
Gene Chromosome Location
Chr.11: 63759892-63768641 on Build GRCh38
Transcripts
NM_001144936.1

SNP Detail

SNP ID Chromosome Location
rs888136763 Chr.11: 63763832-63763832 on Build GRCh38
rs553956173 Chr.11: 63763754-63763754 on Build GRCh38
rs954686228 Chr.11: 63763742-63763742 on Build GRCh38
rs918517625 Chr.11: 63763865-63763865 on Build GRCh38
rs1018275784 Chr.11: 63763814-63763814 on Build GRCh38
rs985576939 Chr.11: 63763813-63763813 on Build GRCh38
rs914046450 Chr.11: 63763769-63763769 on Build GRCh38
rs929838906 Chr.11: 63763740-63763751 on Build GRCh38
rs1018257153 Chr.11: 63763760-63763760 on Build GRCh38
rs752108618 Chr.11: 63763759-63763759 on Build GRCh38
rs1003520439 Chr.11: 63763777-63763777 on Build GRCh38
rs754582462 Chr.11: 63763749-63763757 on Build GRCh38
rs778380840 Chr.11: 63763812-63763814 on Build GRCh38
rs556795894 Chr.11: 63763877-63763877 on Build GRCh38
rs941255967 Chr.11: 63763833-63763833 on Build GRCh38
rs554330843 Chr.11: 63763740-63763740 on Build GRCh38
rs989786394 Chr.11: 63763764-63763764 on Build GRCh38
rs951248664 Chr.11: 63763872-63763874 on Build GRCh38
rs779768091 Chr.11: 63763745-63763746 on Build GRCh38
rs370217341 Chr.11: 63763811-63763811 on Build GRCh38
rs958262987 Chr.11: 63763762-63763762 on Build GRCh38
rs891932786 Chr.11: 63763784-63763784 on Build GRCh38
rs536440387 Chr.11: 63763862-63763862 on Build GRCh38
rs906224377 Chr.11: 63763776-63763776 on Build GRCh38
rs927755297 Chr.11: 63763781-63763781 on Build GRCh38
rs982231709 Chr.11: 63763780-63763780 on Build GRCh38
rs965388599 Chr.11: 63763763-63763763 on Build GRCh38
rs2959886 Chr.11: 63763703-63763703 on Build GRCh38
rs937633387 Chr.11: 63763788-63763788 on Build GRCh38
rs943945187 Chr.11: 63763715-63763715 on Build GRCh38
rs753364149 Chr.11: 63763749-63763760 on Build GRCh38
rs866102626 Chr.11: 63763845-63763845 on Build GRCh38
rs888198027 Chr.11: 63763749-63763749 on Build GRCh38
rs1021680400 Chr.11: 63763804-63763804 on Build GRCh38
rs945570526 Chr.11: 63763778-63763778 on Build GRCh38
rs918392187 Chr.11: 63763726-63763726 on Build GRCh38
rs986107543 Chr.11: 63763745-63763745 on Build GRCh38
rs750270855 Chr.11: 63763737-63763738 on Build GRCh38
rs1010418671 Chr.11: 63763802-63763802 on Build GRCh38

View Full Product Details