Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104588429-104588702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf180
Entrez Gene ID
400258
Gene Name
chromosome 14 open reading frame 180
Gene Aliases
C14orf77, NRAC
UniGene
Hs.153827
Gene Chromosome Location
Chr.14: 104579684-104589847 on Build GRCh38
Transcripts
NM_001008404.2 NM_001286399.1 NM_001286400.1 XM_005267638.3 XM_011536764.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5971052 c.370C>T p.R124W Chr.14: 104588670-104588670 on Build GRCh38
4486773 c.309C>T p.S103S Chr.14: 104588609-104588609 on Build GRCh38
4147851 c.297A>G p.P99P Chr.14: 104588597-104588597 on Build GRCh38
4759740 c.334G>A p.V112I Chr.14: 104588634-104588634 on Build GRCh38
5725021 c.380C>T p.P127L Chr.14: 104588680-104588680 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374323036 Chr.14: 104588681-104588681 on Build GRCh38
rs753361314 Chr.14: 104588643-104588643 on Build GRCh38
rs371187611 Chr.14: 104588540-104588540 on Build GRCh38
rs1031775583 Chr.14: 104588575-104588575 on Build GRCh38
rs756286528 Chr.14: 104588682-104588682 on Build GRCh38
rs758322784 Chr.14: 104588672-104588672 on Build GRCh38
rs111529102 Chr.14: 104588584-104588584 on Build GRCh38
rs374943787 Chr.14: 104588600-104588600 on Build GRCh38
rs753412379 Chr.14: 104588539-104588539 on Build GRCh38
rs12880814 Chr.14: 104588597-104588597 on Build GRCh38
rs774899365 Chr.14: 104588601-104588601 on Build GRCh38
rs955379648 Chr.14: 104588550-104588550 on Build GRCh38
rs113812438 Chr.14: 104588497-104588497 on Build GRCh38
rs906241992 Chr.14: 104588477-104588477 on Build GRCh38
rs73361827 Chr.14: 104588537-104588537 on Build GRCh38
rs565966135 Chr.14: 104588474-104588474 on Build GRCh38
rs868459733 Chr.14: 104588673-104588673 on Build GRCh38
rs892171831 Chr.14: 104588582-104588582 on Build GRCh38
rs1029654490 Chr.14: 104588547-104588547 on Build GRCh38
rs976981412 Chr.14: 104588481-104588481 on Build GRCh38
rs909844837 Chr.14: 104588639-104588639 on Build GRCh38
rs978962396 Chr.14: 104588465-104588465 on Build GRCh38
rs775477825 Chr.14: 104588596-104588596 on Build GRCh38
rs778944156 Chr.14: 104588573-104588573 on Build GRCh38
rs964309917 Chr.14: 104588480-104588480 on Build GRCh38
rs111610399 Chr.14: 104588670-104588670 on Build GRCh38
rs370780261 Chr.14: 104588667-104588667 on Build GRCh38
rs977034607 Chr.14: 104588603-104588603 on Build GRCh38
rs868503379 Chr.14: 104588657-104588657 on Build GRCh38
rs1005725832 Chr.14: 104588530-104588530 on Build GRCh38
rs929952259 Chr.14: 104588488-104588488 on Build GRCh38
rs951676018 Chr.14: 104588631-104588631 on Build GRCh38
rs767921027 Chr.14: 104588606-104588606 on Build GRCh38
rs533562119 Chr.14: 104588666-104588666 on Build GRCh38
rs201205928 Chr.14: 104588538-104588538 on Build GRCh38
rs562143106 Chr.14: 104588583-104588583 on Build GRCh38
rs1046822556 Chr.14: 104588505-104588505 on Build GRCh38
rs535846589 Chr.14: 104588452-104588452 on Build GRCh38
rs983309782 Chr.14: 104588507-104588507 on Build GRCh38
rs370582866 Chr.14: 104588604-104588604 on Build GRCh38
rs868014005 Chr.14: 104588613-104588613 on Build GRCh38
rs534932777 Chr.14: 104588482-104588482 on Build GRCh38
rs377369394 Chr.14: 104588642-104588642 on Build GRCh38
rs555826612 Chr.14: 104588471-104588471 on Build GRCh38
rs373441764 Chr.14: 104588586-104588586 on Build GRCh38
rs773770243 Chr.14: 104588626-104588626 on Build GRCh38
rs968165204 Chr.14: 104588460-104588460 on Build GRCh38
rs557641271 Chr.14: 104588534-104588534 on Build GRCh38
rs115845341 Chr.14: 104588587-104588587 on Build GRCh38
rs573451584 Chr.14: 104588574-104588574 on Build GRCh38
rs541764061 Chr.14: 104588634-104588634 on Build GRCh38

View Full Product Details