Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 40054185-40054438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCOR
Entrez Gene ID
54880
Gene Name
BCL6 corepressor
Gene Aliases
ANOP2, MAA2, MCOPS2
UniGene
Hs.659681
Gene Chromosome Location
Chr.X: 40051246-40177390 on Build GRCh38
Transcripts
NM_001123383.1 NM_001123384.1 NM_001123385.1 NM_017745.5 XM_005272616.1 XM_005272618.3 XM_005272619.3 XM_005272620.3 XM_006724536.3 XM_011543929.2 XM_011543930.1 XM_011543931.2 XM_017029615.1 XM_017029616.1

SNP Detail

SNP ID Chromosome Location
rs768510508 Chr.X: 40054295-40054295 on Build GRCh38
rs758100355 Chr.X: 40054373-40054373 on Build GRCh38
rs367859441 Chr.X: 40054309-40054309 on Build GRCh38
rs898150083 Chr.X: 40054273-40054273 on Build GRCh38
rs779862370 Chr.X: 40054378-40054378 on Build GRCh38
rs767974035 Chr.X: 40054314-40054314 on Build GRCh38
rs749397307 Chr.X: 40054218-40054218 on Build GRCh38
rs772056276 Chr.X: 40054352-40054352 on Build GRCh38
rs752711420 Chr.X: 40054279-40054279 on Build GRCh38
rs754643148 Chr.X: 40054310-40054310 on Build GRCh38
rs771963061 Chr.X: 40054244-40054244 on Build GRCh38
rs1021526994 Chr.X: 40054360-40054360 on Build GRCh38
rs756600306 Chr.X: 40054326-40054326 on Build GRCh38
rs757330389 Chr.X: 40054359-40054359 on Build GRCh38
rs764603328 Chr.X: 40054340-40054340 on Build GRCh38
rs775270452 Chr.X: 40054266-40054266 on Build GRCh38
rs1002602028 Chr.X: 40054269-40054269 on Build GRCh38
rs745593429 Chr.X: 40054239-40054239 on Build GRCh38
rs1033393242 Chr.X: 40054294-40054294 on Build GRCh38
rs776232139 Chr.X: 40054296-40054296 on Build GRCh38
rs761422305 Chr.X: 40054308-40054308 on Build GRCh38
rs1001793293 Chr.X: 40054284-40054284 on Build GRCh38
rs112054628 Chr.X: 40054402-40054402 on Build GRCh38
rs201189214 Chr.X: 40054231-40054231 on Build GRCh38
rs202010863 Chr.X: 40054213-40054213 on Build GRCh38
rs746154364 Chr.X: 40054369-40054369 on Build GRCh38
rs375312850 Chr.X: 40054287-40054287 on Build GRCh38
rs771089228 Chr.X: 40054229-40054229 on Build GRCh38
rs753112982 Chr.X: 40054315-40054315 on Build GRCh38
rs1044537268 Chr.X: 40054222-40054222 on Build GRCh38
rs371551080 Chr.X: 40054368-40054368 on Build GRCh38
rs753853933 Chr.X: 40054358-40054358 on Build GRCh38

View Full Product Details