Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39744177-39744402 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTHD1
Entrez Gene ID
150350
Gene Name
ENTH domain containing 1
Gene Aliases
CACNA1I, dJ370M22.3
UniGene
Hs.474869
Gene Chromosome Location
Chr.22: 39743044-39893884 on Build GRCh38
Transcripts
NM_152512.3 XM_006724149.3 XM_006724150.3 XM_006724151.1 XM_011529927.2 XM_011529928.2 XM_011529930.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1714409 c.1313C>T p.S438L Chr.22: 39744190-39744190 on Build GRCh38
5859424 c.1256C>T p.P419L Chr.22: 39744247-39744247 on Build GRCh38
3554753 c.1223C>T p.S408L Chr.22: 39744280-39744280 on Build GRCh38
6276454 c.1315C>A p.P439T Chr.22: 39744188-39744188 on Build GRCh38
3554752 c.1262C>T p.S421F Chr.22: 39744241-39744241 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767590468 Chr.22: 39744285-39744285 on Build GRCh38
rs765680600 Chr.22: 39744319-39744319 on Build GRCh38
rs868164759 Chr.22: 39744281-39744281 on Build GRCh38
rs752651560 Chr.22: 39744241-39744241 on Build GRCh38
rs374392116 Chr.22: 39744244-39744244 on Build GRCh38
rs191007110 Chr.22: 39744294-39744294 on Build GRCh38
rs758936453 Chr.22: 39744324-39744324 on Build GRCh38
rs780779007 Chr.22: 39744328-39744328 on Build GRCh38
rs200008989 Chr.22: 39744298-39744298 on Build GRCh38
rs762780840 Chr.22: 39744239-39744239 on Build GRCh38
rs775390948 Chr.22: 39744292-39744292 on Build GRCh38
rs771713637 Chr.22: 39744356-39744356 on Build GRCh38
rs755622430 Chr.22: 39744334-39744334 on Build GRCh38
rs759413013 Chr.22: 39744259-39744259 on Build GRCh38
rs747664310 Chr.22: 39744332-39744332 on Build GRCh38
rs904668138 Chr.22: 39744366-39744366 on Build GRCh38
rs769284855 Chr.22: 39744214-39744214 on Build GRCh38
rs757758541 Chr.22: 39744307-39744307 on Build GRCh38
rs1039634295 Chr.22: 39744330-39744330 on Build GRCh38
rs918254917 Chr.22: 39744220-39744220 on Build GRCh38
rs115302874 Chr.22: 39744238-39744238 on Build GRCh38
rs115673469 Chr.22: 39744223-39744223 on Build GRCh38
rs750977073 Chr.22: 39744323-39744323 on Build GRCh38
rs943589573 Chr.22: 39744299-39744299 on Build GRCh38

View Full Product Details