Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 69282624-69282885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC125
Entrez Gene ID
202243
Gene Name
coiled-coil domain containing 125
Gene Aliases
KENAE
UniGene
Hs.654850
Gene Chromosome Location
Chr.5: 69273087-69333069 on Build GRCh38
Transcripts
NM_001297696.1 NM_001297697.1 NM_176816.4 XM_005248458.4 XM_005248459.4 XM_005248461.3 XM_005248463.3 XM_005248464.3 XM_006714569.3 XM_006714570.3 XM_011543255.2 XM_011543256.2 XM_011543257.2 XM_011543258.2 XM_011543259.2 XM_017009206.1 XM_017009207.1 XM_017009208.1 XM_017009209.1 XM_017009210.1 XM_017009211.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1069715 c.1502G>T p.R501I Chr.5: 69282763-69282763 on Build GRCh38
279617 c.1477G>T p.D493Y Chr.5: 69282788-69282788 on Build GRCh38
5806288 c.1494T>C p.T498T Chr.5: 69282771-69282771 on Build GRCh38
2155472 c.1451T>C p.I484T Chr.5: 69282814-69282814 on Build GRCh38
6584503 c.1471C>G p.Q491E Chr.5: 69282794-69282794 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766365951 Chr.5: 69282682-69282682 on Build GRCh38
rs747848919 Chr.5: 69282756-69282756 on Build GRCh38
rs765432478 Chr.5: 69282692-69282692 on Build GRCh38
rs370270501 Chr.5: 69282857-69282857 on Build GRCh38
rs757824120 Chr.5: 69282839-69282840 on Build GRCh38
rs571097830 Chr.5: 69282752-69282752 on Build GRCh38
rs777007275 Chr.5: 69282792-69282792 on Build GRCh38
rs772395128 Chr.5: 69282761-69282761 on Build GRCh38
rs934143856 Chr.5: 69282711-69282711 on Build GRCh38
rs771411085 Chr.5: 69282778-69282778 on Build GRCh38
rs767411339 Chr.5: 69282845-69282845 on Build GRCh38
rs372457894 Chr.5: 69282710-69282710 on Build GRCh38
rs186008572 Chr.5: 69282687-69282687 on Build GRCh38
rs370091238 Chr.5: 69282688-69282688 on Build GRCh38
rs763303289 Chr.5: 69282843-69282843 on Build GRCh38
rs760623546 Chr.5: 69282698-69282699 on Build GRCh38
rs749372673 Chr.5: 69282726-69282726 on Build GRCh38
rs747457653 Chr.5: 69282777-69282777 on Build GRCh38
rs759618555 Chr.5: 69282796-69282796 on Build GRCh38
rs559271914 Chr.5: 69282742-69282742 on Build GRCh38
rs757031007 Chr.5: 69282736-69282738 on Build GRCh38
rs144635477 Chr.5: 69282775-69282775 on Build GRCh38
rs56680316 Chr.5: 69282679-69282679 on Build GRCh38
rs761211727 Chr.5: 69282681-69282681 on Build GRCh38
rs1032003618 Chr.5: 69282844-69282844 on Build GRCh38
rs775829106 Chr.5: 69282842-69282842 on Build GRCh38
rs753630915 Chr.5: 69282730-69282731 on Build GRCh38
rs889531946 Chr.5: 69282741-69282741 on Build GRCh38
rs775315552 Chr.5: 69282694-69282696 on Build GRCh38
rs1008030732 Chr.5: 69282698-69282698 on Build GRCh38
rs757065653 Chr.5: 69282715-69282715 on Build GRCh38
rs752661627 Chr.5: 69282768-69282768 on Build GRCh38
rs374598741 Chr.5: 69282813-69282813 on Build GRCh38
rs972945937 Chr.5: 69282705-69282705 on Build GRCh38
rs1043417788 Chr.5: 69282825-69282825 on Build GRCh38
rs770987300 Chr.5: 69282728-69282728 on Build GRCh38
rs748768744 Chr.5: 69282746-69282746 on Build GRCh38
rs903479845 Chr.5: 69282855-69282855 on Build GRCh38
rs767466170 Chr.5: 69282727-69282727 on Build GRCh38
rs764977669 Chr.5: 69282739-69282740 on Build GRCh38
rs139850727 Chr.5: 69282820-69282820 on Build GRCh38
rs754479533 Chr.5: 69282748-69282748 on Build GRCh38
rs201030560 Chr.5: 69282841-69282841 on Build GRCh38
rs749994635 Chr.5: 69282757-69282758 on Build GRCh38

View Full Product Details