Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 108752806-108753067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETNPPL
Entrez Gene ID
64850
Gene Name
ethanolamine-phosphate phospho-lyase
Gene Aliases
AGXT2L1
UniGene
Hs.106576
Gene Chromosome Location
Chr.4: 108742040-108763180 on Build GRCh38
Transcripts
NM_001146590.1 NM_001146627.1 NM_031279.3 NR_027474.1 NR_027475.1 XM_006714290.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5918335 c.526G>A p.G176R Chr.4: 108752987-108752987 on Build GRCh38
3916925 c.527G>A p.G176E Chr.4: 108752986-108752986 on Build GRCh38
6442776 c.592G>C p.E198Q Chr.4: 108752921-108752921 on Build GRCh38
4638609 c.579G>T p.V193V Chr.4: 108752934-108752934 on Build GRCh38
4121252 c.519T>G p.T173T Chr.4: 108752994-108752994 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781118542 Chr.4: 108752934-108752934 on Build GRCh38
rs148598163 Chr.4: 108752940-108752940 on Build GRCh38
rs761403913 Chr.4: 108752979-108752979 on Build GRCh38
rs772650876 Chr.4: 108752972-108752972 on Build GRCh38
rs568987227 Chr.4: 108753028-108753028 on Build GRCh38
rs762467940 Chr.4: 108753018-108753018 on Build GRCh38
rs764969848 Chr.4: 108752880-108752880 on Build GRCh38
rs374710518 Chr.4: 108752908-108752908 on Build GRCh38
rs201217309 Chr.4: 108752987-108752987 on Build GRCh38
rs138316049 Chr.4: 108753033-108753033 on Build GRCh38
rs992161853 Chr.4: 108752966-108752966 on Build GRCh38
rs759430382 Chr.4: 108753029-108753029 on Build GRCh38
rs146141629 Chr.4: 108752948-108752948 on Build GRCh38
rs371747173 Chr.4: 108753022-108753022 on Build GRCh38
rs769469536 Chr.4: 108753000-108753000 on Build GRCh38
rs750168429 Chr.4: 108752881-108752881 on Build GRCh38
rs372065522 Chr.4: 108752965-108752965 on Build GRCh38
rs777936791 Chr.4: 108752953-108752953 on Build GRCh38
rs917918165 Chr.4: 108752971-108752971 on Build GRCh38
rs147464331 Chr.4: 108752858-108752858 on Build GRCh38
rs368089283 Chr.4: 108753005-108753005 on Build GRCh38
rs530138972 Chr.4: 108752851-108752851 on Build GRCh38
rs1377210 Chr.4: 108752960-108752960 on Build GRCh38
rs540319041 Chr.4: 108752857-108752857 on Build GRCh38
rs201996006 Chr.4: 108753027-108753027 on Build GRCh38
rs929410500 Chr.4: 108753007-108753007 on Build GRCh38
rs375012393 Chr.4: 108752974-108752974 on Build GRCh38
rs751396181 Chr.4: 108752897-108752897 on Build GRCh38
rs776215719 Chr.4: 108752862-108752862 on Build GRCh38
rs979234834 Chr.4: 108752876-108752876 on Build GRCh38
rs774899369 Chr.4: 108752853-108752853 on Build GRCh38
rs766141211 Chr.4: 108752894-108752894 on Build GRCh38
rs761424100 Chr.4: 108752870-108752870 on Build GRCh38
rs142970921 Chr.4: 108752946-108752946 on Build GRCh38
rs185770114 Chr.4: 108752955-108752955 on Build GRCh38

View Full Product Details