Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 75379349-75379574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL4A3BP
Entrez Gene ID
10087
Gene Name
collagen type IV alpha 3 binding protein
Gene Aliases
CERT, CERTL, GPBP, MRD34, STARD11
UniGene
Hs.270437
Gene Chromosome Location
Chr.5: 75368486-75511981 on Build GRCh38
Transcripts
NM_001130105.1 NM_005713.2 NM_031361.2 XM_006714513.2 XM_011543090.2 XM_011543091.2 XM_017008919.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6332483 c.1748-7C>T p.? Chr.5: 75379480-75379480 on Build GRCh38
4621194 c.2188T>C p.Y730H Chr.5: 75379417-75379417 on Build GRCh38
6359297 c.1856G>T p.G619V Chr.5: 75379365-75379365 on Build GRCh38
1201909 c.1798C>T p.R600* Chr.5: 75379423-75379423 on Build GRCh38
5432210 c.2238A>T p.A746A Chr.5: 75379367-75379367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371046717 Chr.5: 75379421-75379421 on Build GRCh38
rs371299966 Chr.5: 75379523-75379523 on Build GRCh38
rs768793437 Chr.5: 75379401-75379401 on Build GRCh38
rs759226067 Chr.5: 75379460-75379460 on Build GRCh38
rs750059205 Chr.5: 75379508-75379509 on Build GRCh38
rs55882089 Chr.5: 75379425-75379425 on Build GRCh38
rs376382310 Chr.5: 75379384-75379384 on Build GRCh38
rs766212065 Chr.5: 75379441-75379441 on Build GRCh38
rs763174978 Chr.5: 75379416-75379416 on Build GRCh38
rs373547173 Chr.5: 75379388-75379388 on Build GRCh38
rs565479126 Chr.5: 75379385-75379385 on Build GRCh38
rs772106628 Chr.5: 75379417-75379417 on Build GRCh38
rs1054243983 Chr.5: 75379541-75379541 on Build GRCh38
rs1013864471 Chr.5: 75379423-75379423 on Build GRCh38
rs753368457 Chr.5: 75379442-75379442 on Build GRCh38
rs190220472 Chr.5: 75379477-75379477 on Build GRCh38
rs781220540 Chr.5: 75379376-75379376 on Build GRCh38
rs950732236 Chr.5: 75379456-75379456 on Build GRCh38

View Full Product Details