Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 25904889-25905150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APP
Entrez Gene ID
351
Gene Name
amyloid beta precursor protein
Gene Aliases
AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN-II, PN2
UniGene
Hs.434980
Gene Chromosome Location
Chr.21: 25880550-26171128 on Build GRCh38
Transcripts
NM_000484.3 NM_001136016.3 NM_001136129.2 NM_001136130.2 NM_001136131.2 NM_001204301.1 NM_001204302.1 NM_001204303.1 NM_201413.2 NM_201414.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2841337 c.1962A>G p.P654P Chr.21: 25905025-25905025 on Build GRCh38
6565424 c.1912G>C p.E638Q Chr.21: 25905075-25905075 on Build GRCh38
1029639 c.1954A>G p.T652A Chr.21: 25905033-25905033 on Build GRCh38
6355160 c.1938C>T p.A646A Chr.21: 25905049-25905049 on Build GRCh38
5805230 c.1958G>A p.R653Q Chr.21: 25905029-25905029 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764881231 Chr.21: 25905113-25905114 on Build GRCh38
rs145273675 Chr.21: 25905049-25905049 on Build GRCh38
rs1037461053 Chr.21: 25904936-25904936 on Build GRCh38
rs1006102441 Chr.21: 25904957-25904957 on Build GRCh38
rs201038323 Chr.21: 25905046-25905046 on Build GRCh38
rs45558740 Chr.21: 25904966-25904966 on Build GRCh38
rs201882791 Chr.21: 25905080-25905080 on Build GRCh38
rs780032914 Chr.21: 25905085-25905085 on Build GRCh38
rs771520531 Chr.21: 25905002-25905002 on Build GRCh38
rs778128146 Chr.21: 25904980-25904980 on Build GRCh38
rs199932473 Chr.21: 25905031-25905031 on Build GRCh38
rs763165305 Chr.21: 25905020-25905020 on Build GRCh38
rs151010236 Chr.21: 25905059-25905059 on Build GRCh38
rs749023520 Chr.21: 25904981-25904981 on Build GRCh38
rs760100945 Chr.21: 25905034-25905034 on Build GRCh38
rs200785502 Chr.21: 25904991-25904991 on Build GRCh38
rs1040963302 Chr.21: 25905103-25905103 on Build GRCh38
rs745838621 Chr.21: 25904997-25904997 on Build GRCh38
rs768182065 Chr.21: 25905045-25905045 on Build GRCh38
rs952919904 Chr.21: 25904945-25904945 on Build GRCh38
rs961647379 Chr.21: 25904977-25904977 on Build GRCh38
rs200287355 Chr.21: 25904975-25904975 on Build GRCh38
rs200910966 Chr.21: 25905006-25905006 on Build GRCh38
rs778792696 Chr.21: 25905061-25905061 on Build GRCh38
rs370979414 Chr.21: 25905030-25905030 on Build GRCh38
rs368159818 Chr.21: 25905048-25905048 on Build GRCh38
rs538528103 Chr.21: 25904922-25904922 on Build GRCh38
rs765412723 Chr.21: 25905044-25905044 on Build GRCh38
rs191673387 Chr.21: 25904976-25904976 on Build GRCh38
rs199696263 Chr.21: 25904974-25904974 on Build GRCh38
rs763648992 Chr.21: 25905013-25905013 on Build GRCh38
rs45467193 Chr.21: 25904928-25904928 on Build GRCh38
rs574536250 Chr.21: 25904982-25904982 on Build GRCh38
rs182142845 Chr.21: 25905130-25905130 on Build GRCh38
rs771964280 Chr.21: 25905071-25905071 on Build GRCh38
rs768221786 Chr.21: 25905101-25905101 on Build GRCh38
rs760218235 Chr.21: 25905012-25905012 on Build GRCh38
rs199707780 Chr.21: 25904994-25904994 on Build GRCh38
rs201884382 Chr.21: 25905014-25905014 on Build GRCh38
rs201706747 Chr.21: 25904993-25904993 on Build GRCh38
rs745710188 Chr.21: 25905067-25905067 on Build GRCh38
rs199540835 Chr.21: 25904983-25904983 on Build GRCh38
rs991217758 Chr.21: 25905017-25905017 on Build GRCh38
rs200115611 Chr.21: 25905095-25905095 on Build GRCh38
rs201099956 Chr.21: 25905097-25905097 on Build GRCh38
rs776399216 Chr.21: 25905019-25905019 on Build GRCh38
rs982322907 Chr.21: 25904978-25904978 on Build GRCh38
rs542928784 Chr.21: 25905102-25905105 on Build GRCh38

View Full Product Details