Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62603092-62603317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EML3
Entrez Gene ID
256364
Gene Name
echinoderm microtubule associated protein like 3
Gene Aliases
ELP95
UniGene
Hs.379785
Gene Chromosome Location
Chr.11: 62602218-62613020 on Build GRCh38
Transcripts
NM_001300793.1 NM_001300794.1 NM_153265.2 XM_005273878.3 XM_006718489.3 XM_006718490.3 XM_006718491.3 XM_011544896.2 XM_011544897.2 XM_017017480.1 XM_017017481.1 XM_017017482.1 XM_017017483.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6292035 c.2258-8C>T p.? Chr.11: 62603255-62603255 on Build GRCh38
3809835 c.2289G>T p.K763N Chr.11: 62603216-62603216 on Build GRCh38
929724 c.2270G>A p.G757E Chr.11: 62603235-62603235 on Build GRCh38
6653812 c.2298T>C p.Y766Y Chr.11: 62603207-62603207 on Build GRCh38
3809836 c.2289G>T p.K763N Chr.11: 62603216-62603216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749854968 Chr.11: 62603257-62603257 on Build GRCh38
rs745489931 Chr.11: 62603289-62603289 on Build GRCh38
rs757439722 Chr.11: 62603242-62603242 on Build GRCh38
rs776409158 Chr.11: 62603268-62603268 on Build GRCh38
rs762850607 Chr.11: 62603231-62603231 on Build GRCh38
rs764194625 Chr.11: 62603234-62603234 on Build GRCh38
rs750608225 Chr.11: 62603244-62603244 on Build GRCh38
rs997630807 Chr.11: 62603138-62603138 on Build GRCh38
rs777451133 Chr.11: 62603261-62603261 on Build GRCh38
rs914307942 Chr.11: 62603287-62603287 on Build GRCh38
rs1054080608 Chr.11: 62603278-62603278 on Build GRCh38
rs759307596 Chr.11: 62603204-62603204 on Build GRCh38
rs183273900 Chr.11: 62603253-62603253 on Build GRCh38
rs909444158 Chr.11: 62603252-62603252 on Build GRCh38
rs746998497 Chr.11: 62603135-62603137 on Build GRCh38
rs772818496 Chr.11: 62603178-62603178 on Build GRCh38
rs776301213 Chr.11: 62603200-62603200 on Build GRCh38
rs746473202 Chr.11: 62603265-62603265 on Build GRCh38
rs760436124 Chr.11: 62603184-62603184 on Build GRCh38
rs190938643 Chr.11: 62603192-62603192 on Build GRCh38
rs780777645 Chr.11: 62603143-62603143 on Build GRCh38
rs950352101 Chr.11: 62603194-62603194 on Build GRCh38
rs371861729 Chr.11: 62603238-62603238 on Build GRCh38
rs771757249 Chr.11: 62603167-62603167 on Build GRCh38
rs770491157 Chr.11: 62603266-62603266 on Build GRCh38
rs764935495 Chr.11: 62603226-62603226 on Build GRCh38
rs755505749 Chr.11: 62603259-62603259 on Build GRCh38
rs752688797 Chr.11: 62603228-62603228 on Build GRCh38
rs746187522 Chr.11: 62603119-62603119 on Build GRCh38
rs770990530 Chr.11: 62603139-62603140 on Build GRCh38
rs749804407 Chr.11: 62603158-62603158 on Build GRCh38
rs770210685 Chr.11: 62603125-62603125 on Build GRCh38
rs527676386 Chr.11: 62603243-62603243 on Build GRCh38
rs5792263 Chr.11: 62603245-62603245 on Build GRCh38

View Full Product Details