Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 84410847-84411108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNASE2B
Entrez Gene ID
58511
Gene Name
deoxyribonuclease II beta
Gene Aliases
DLAD
UniGene
Hs.129142
Gene Chromosome Location
Chr.1: 84398532-84415895 on Build GRCh38
Transcripts
NM_021233.2 NM_058248.1 XM_011541878.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1602715 c.455C>T p.P152L Chr.1: 84410907-84410907 on Build GRCh38
4496460 c.475C>T p.P159S Chr.1: 84410927-84410927 on Build GRCh38
7298086 c.516A>G p.I172M Chr.1: 84410968-84410968 on Build GRCh38
5506616 c.437C>A p.P146H Chr.1: 84410889-84410889 on Build GRCh38
6322240 c.396G>T p.L132L Chr.1: 84410848-84410848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867997727 Chr.1: 84411062-84411062 on Build GRCh38
rs146489254 Chr.1: 84411052-84411052 on Build GRCh38
rs756099350 Chr.1: 84410942-84410942 on Build GRCh38
rs756187232 Chr.1: 84411029-84411029 on Build GRCh38
rs779939965 Chr.1: 84410943-84410943 on Build GRCh38
rs759806353 Chr.1: 84410993-84410993 on Build GRCh38
rs768417609 Chr.1: 84410957-84410957 on Build GRCh38
rs368419588 Chr.1: 84410976-84410976 on Build GRCh38
rs753683784 Chr.1: 84411037-84411037 on Build GRCh38
rs763150255 Chr.1: 84411014-84411014 on Build GRCh38
rs747665428 Chr.1: 84410968-84410968 on Build GRCh38
rs771308390 Chr.1: 84410989-84410989 on Build GRCh38
rs773644888 Chr.1: 84410897-84410897 on Build GRCh38
rs778375234 Chr.1: 84410966-84410966 on Build GRCh38
rs937155369 Chr.1: 84410958-84410958 on Build GRCh38
rs149919241 Chr.1: 84410997-84410997 on Build GRCh38
rs752794024 Chr.1: 84411007-84411007 on Build GRCh38
rs768028542 Chr.1: 84410884-84410884 on Build GRCh38
rs373925401 Chr.1: 84410931-84410931 on Build GRCh38
rs67734584 Chr.1: 84411029-84411030 on Build GRCh38
rs3105610 Chr.1: 84411017-84411017 on Build GRCh38
rs766678712 Chr.1: 84410903-84410903 on Build GRCh38
rs927051150 Chr.1: 84410955-84410955 on Build GRCh38
rs944383306 Chr.1: 84410886-84410886 on Build GRCh38
rs3216015 Chr.1: 84411028-84411029 on Build GRCh38
rs751967935 Chr.1: 84410907-84410907 on Build GRCh38
rs893046549 Chr.1: 84410975-84410975 on Build GRCh38
rs78247834 Chr.1: 84411027-84411027 on Build GRCh38
rs370520060 Chr.1: 84410913-84410913 on Build GRCh38
rs747551768 Chr.1: 84411019-84411020 on Build GRCh38
rs758492822 Chr.1: 84410926-84410926 on Build GRCh38
rs765373252 Chr.1: 84410912-84410912 on Build GRCh38
rs761215984 Chr.1: 84410983-84410983 on Build GRCh38
rs368268321 Chr.1: 84410908-84410908 on Build GRCh38
rs199729041 Chr.1: 84410877-84410877 on Build GRCh38
rs745752643 Chr.1: 84410938-84410938 on Build GRCh38
rs180791234 Chr.1: 84411039-84411039 on Build GRCh38
rs761303620 Chr.1: 84410880-84410880 on Build GRCh38
rs771533004 Chr.1: 84410973-84410973 on Build GRCh38
rs199881802 Chr.1: 84410872-84410872 on Build GRCh38
rs781277278 Chr.1: 84410930-84410930 on Build GRCh38
rs780787182 Chr.1: 84410952-84410953 on Build GRCh38
rs1506700 Chr.1: 84411048-84411048 on Build GRCh38
rs57249159 Chr.1: 84410902-84410902 on Build GRCh38
rs749031735 Chr.1: 84410956-84410956 on Build GRCh38
rs779846457 Chr.1: 84411033-84411033 on Build GRCh38
rs199974554 Chr.1: 84411028-84411028 on Build GRCh38
rs752878541 Chr.1: 84410921-84410921 on Build GRCh38

View Full Product Details