Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109923168-109923441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSF1
Entrez Gene ID
1435
Gene Name
colony stimulating factor 1
Gene Aliases
CSF-1, MCSF
UniGene
Hs.173894
Gene Chromosome Location
Chr.1: 109910506-109930994 on Build GRCh38
Transcripts
NM_000757.5 NM_172210.2 NM_172211.3 NM_172212.2 XM_017000369.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6057601 c.688G>A p.E230K Chr.1: 109923309-109923309 on Build GRCh38
4503419 c.639C>T p.P213P Chr.1: 109923260-109923260 on Build GRCh38
4791723 c.697T>G p.S233A Chr.1: 109923318-109923318 on Build GRCh38
7263763 c.750G>A p.K250K Chr.1: 109923371-109923371 on Build GRCh38
3862148 c.629C>T p.P210L Chr.1: 109923250-109923250 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527845264 Chr.1: 109923405-109923405 on Build GRCh38
rs562274652 Chr.1: 109923349-109923349 on Build GRCh38
rs764752158 Chr.1: 109923339-109923339 on Build GRCh38
rs548258030 Chr.1: 109923319-109923319 on Build GRCh38
rs147308330 Chr.1: 109923226-109923226 on Build GRCh38
rs752363763 Chr.1: 109923236-109923236 on Build GRCh38
rs756637398 Chr.1: 109923369-109923369 on Build GRCh38
rs140214194 Chr.1: 109923212-109923212 on Build GRCh38
rs111387080 Chr.1: 109923248-109923248 on Build GRCh38
rs774283172 Chr.1: 109923200-109923200 on Build GRCh38
rs148944210 Chr.1: 109923260-109923260 on Build GRCh38
rs755112511 Chr.1: 109923268-109923268 on Build GRCh38
rs267597915 Chr.1: 109923274-109923274 on Build GRCh38
rs369018338 Chr.1: 109923354-109923354 on Build GRCh38
rs202063964 Chr.1: 109923214-109923214 on Build GRCh38
rs767573181 Chr.1: 109923421-109923421 on Build GRCh38
rs113687620 Chr.1: 109923265-109923265 on Build GRCh38
rs758272243 Chr.1: 109923307-109923307 on Build GRCh38
rs779161150 Chr.1: 109923388-109923388 on Build GRCh38
rs868315527 Chr.1: 109923406-109923406 on Build GRCh38
rs752931255 Chr.1: 109923397-109923397 on Build GRCh38
rs137985559 Chr.1: 109923409-109923409 on Build GRCh38
rs148259638 Chr.1: 109923329-109923329 on Build GRCh38
rs750913335 Chr.1: 109923255-109923255 on Build GRCh38
rs769056430 Chr.1: 109923191-109923191 on Build GRCh38
rs771856655 Chr.1: 109923304-109923304 on Build GRCh38
rs200796309 Chr.1: 109923313-109923313 on Build GRCh38
rs760559037 Chr.1: 109923217-109923217 on Build GRCh38
rs776179599 Chr.1: 109923220-109923220 on Build GRCh38
rs771774366 Chr.1: 109923213-109923213 on Build GRCh38
rs954483459 Chr.1: 109923243-109923243 on Build GRCh38
rs748400342 Chr.1: 109923287-109923287 on Build GRCh38
rs770844361 Chr.1: 109923320-109923320 on Build GRCh38
rs267597914 Chr.1: 109923269-109923269 on Build GRCh38
rs764807646 Chr.1: 109923227-109923227 on Build GRCh38
rs931322068 Chr.1: 109923345-109923345 on Build GRCh38
rs779852469 Chr.1: 109923309-109923309 on Build GRCh38
rs373284627 Chr.1: 109923410-109923410 on Build GRCh38
rs754408495 Chr.1: 109923376-109923376 on Build GRCh38
rs199934990 Chr.1: 109923343-109923343 on Build GRCh38
rs767804510 Chr.1: 109923254-109923254 on Build GRCh38
rs750809815 Chr.1: 109923364-109923364 on Build GRCh38
rs780266797 Chr.1: 109923263-109923263 on Build GRCh38
rs765879124 Chr.1: 109923218-109923218 on Build GRCh38
rs756629627 Chr.1: 109923258-109923258 on Build GRCh38
rs149810852 Chr.1: 109923252-109923252 on Build GRCh38
rs755057012 Chr.1: 109923382-109923382 on Build GRCh38
rs199659620 Chr.1: 109923201-109923201 on Build GRCh38
rs887172482 Chr.1: 109923378-109923378 on Build GRCh38
rs776556568 Chr.1: 109923323-109923323 on Build GRCh38
rs766926552 Chr.1: 109923375-109923375 on Build GRCh38

View Full Product Details