Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37840004-37840267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD54
Entrez Gene ID
129138
Gene Name
ankyrin repeat domain 54
Gene Aliases
LIAR
UniGene
Hs.135259
Gene Chromosome Location
Chr.22: 37830854-37848994 on Build GRCh38
Transcripts
NM_138797.2 NR_036556.1 XM_006724136.1 XM_006724137.1 XM_011529877.2 XM_011529878.2 XM_011529879.2 XM_011529880.2 XM_011529882.2 XM_011529883.2 XM_011529884.2 XM_017028590.1 XM_017028591.1 XM_017028593.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4413834 c.329C>G p.A110G Chr.22: 37840234-37840234 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs377730696 Chr.22: 37840049-37840052 on Build GRCh38
rs1038077616 Chr.22: 37840091-37840091 on Build GRCh38
rs753617341 Chr.22: 37840172-37840172 on Build GRCh38
rs189860207 Chr.22: 37840207-37840207 on Build GRCh38
rs979544003 Chr.22: 37840186-37840186 on Build GRCh38
rs767619849 Chr.22: 37840151-37840151 on Build GRCh38
rs773036198 Chr.22: 37840154-37840154 on Build GRCh38
rs915616195 Chr.22: 37840168-37840168 on Build GRCh38
rs768978927 Chr.22: 37840140-37840140 on Build GRCh38
rs1045383079 Chr.22: 37840045-37840045 on Build GRCh38
rs202243316 Chr.22: 37840156-37840156 on Build GRCh38
rs905498848 Chr.22: 37840081-37840081 on Build GRCh38
rs994329174 Chr.22: 37840097-37840097 on Build GRCh38
rs949980728 Chr.22: 37840030-37840030 on Build GRCh38
rs564209234 Chr.22: 37840095-37840095 on Build GRCh38
rs757921384 Chr.22: 37840201-37840201 on Build GRCh38
rs768222449 Chr.22: 37840129-37840129 on Build GRCh38
rs769064788 Chr.22: 37840237-37840237 on Build GRCh38
rs764868784 Chr.22: 37840181-37840181 on Build GRCh38
rs780715015 Chr.22: 37840212-37840212 on Build GRCh38
rs111377541 Chr.22: 37840195-37840195 on Build GRCh38
rs967646592 Chr.22: 37840076-37840076 on Build GRCh38
rs751423899 Chr.22: 37840116-37840116 on Build GRCh38
rs531171205 Chr.22: 37840142-37840142 on Build GRCh38
rs561938325 Chr.22: 37840219-37840219 on Build GRCh38
rs150780619 Chr.22: 37840227-37840227 on Build GRCh38
rs925714178 Chr.22: 37840127-37840127 on Build GRCh38
rs754670945 Chr.22: 37840176-37840176 on Build GRCh38
rs375146500 Chr.22: 37840138-37840138 on Build GRCh38
rs777274062 Chr.22: 37840206-37840206 on Build GRCh38
rs369399254 Chr.22: 37840224-37840224 on Build GRCh38
rs1018288990 Chr.22: 37840162-37840162 on Build GRCh38
rs955662114 Chr.22: 37840131-37840131 on Build GRCh38
rs779263534 Chr.22: 37840241-37840241 on Build GRCh38
rs1001279989 Chr.22: 37840087-37840087 on Build GRCh38
rs780043733 Chr.22: 37840115-37840115 on Build GRCh38

View Full Product Details