Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 34197669-34197942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf94
Entrez Gene ID
84970
Gene Name
chromosome 1 open reading frame 94
Gene Aliases
-
UniGene
Hs.194610
Gene Chromosome Location
Chr.1: 34166883-34219131 on Build GRCh38
Transcripts
NM_001134734.1 NM_032884.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5559263 c.370C>T p.L124F Chr.1: 34197844-34197844 on Build GRCh38
228786 c.253T>C p.F85L Chr.1: 34197727-34197727 on Build GRCh38
6063301 c.883C>A p.R295R Chr.1: 34197787-34197787 on Build GRCh38
6126502 c.199A>G p.K67E Chr.1: 34197673-34197673 on Build GRCh38
5342355 c.919C>T p.L307F Chr.1: 34197823-34197823 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779510640 Chr.1: 34197768-34197771 on Build GRCh38
rs551996670 Chr.1: 34197695-34197695 on Build GRCh38
rs890507539 Chr.1: 34197704-34197704 on Build GRCh38
rs776385463 Chr.1: 34197834-34197834 on Build GRCh38
rs778648381 Chr.1: 34197774-34197774 on Build GRCh38
rs556951025 Chr.1: 34197805-34197805 on Build GRCh38
rs148899786 Chr.1: 34197902-34197902 on Build GRCh38
rs748261972 Chr.1: 34197787-34197787 on Build GRCh38
rs776024768 Chr.1: 34197919-34197919 on Build GRCh38
rs775804031 Chr.1: 34197815-34197815 on Build GRCh38
rs982589175 Chr.1: 34197866-34197866 on Build GRCh38
rs140503394 Chr.1: 34197873-34197873 on Build GRCh38
rs760038036 Chr.1: 34197748-34197748 on Build GRCh38
rs752285731 Chr.1: 34197747-34197747 on Build GRCh38
rs545905362 Chr.1: 34197909-34197909 on Build GRCh38
rs750407248 Chr.1: 34197775-34197775 on Build GRCh38
rs372313446 Chr.1: 34197702-34197702 on Build GRCh38
rs1056169685 Chr.1: 34197889-34197889 on Build GRCh38
rs1414474 Chr.1: 34197810-34197810 on Build GRCh38
rs535454453 Chr.1: 34197803-34197803 on Build GRCh38
rs757586827 Chr.1: 34197892-34197892 on Build GRCh38
rs367889351 Chr.1: 34197802-34197802 on Build GRCh38
rs148742081 Chr.1: 34197739-34197739 on Build GRCh38
rs749335287 Chr.1: 34197885-34197885 on Build GRCh38
rs771405561 Chr.1: 34197735-34197735 on Build GRCh38
rs200445096 Chr.1: 34197788-34197788 on Build GRCh38
rs147621215 Chr.1: 34197874-34197874 on Build GRCh38
rs184707165 Chr.1: 34197738-34197738 on Build GRCh38
rs1017953867 Chr.1: 34197759-34197759 on Build GRCh38
rs775041554 Chr.1: 34197736-34197736 on Build GRCh38
rs750247841 Chr.1: 34197858-34197858 on Build GRCh38
rs768260688 Chr.1: 34197918-34197918 on Build GRCh38
rs145927354 Chr.1: 34197881-34197881 on Build GRCh38
rs753421284 Chr.1: 34197753-34197753 on Build GRCh38
rs757109947 Chr.1: 34197755-34197755 on Build GRCh38
rs768505494 Chr.1: 34197869-34197869 on Build GRCh38
rs761432197 Chr.1: 34197845-34197845 on Build GRCh38
rs758119685 Chr.1: 34197862-34197862 on Build GRCh38
rs772554672 Chr.1: 34197903-34197903 on Build GRCh38
rs1045853410 Chr.1: 34197907-34197907 on Build GRCh38
rs747441377 Chr.1: 34197912-34197912 on Build GRCh38
rs778111912 Chr.1: 34197796-34197796 on Build GRCh38
rs763509116 Chr.1: 34197825-34197825 on Build GRCh38
rs768090484 Chr.1: 34197743-34197743 on Build GRCh38
rs748126071 Chr.1: 34197698-34197698 on Build GRCh38
rs866552624 Chr.1: 34197833-34197833 on Build GRCh38
rs555746997 Chr.1: 34197751-34197751 on Build GRCh38
rs773238507 Chr.1: 34197719-34197719 on Build GRCh38
rs770082695 Chr.1: 34197703-34197703 on Build GRCh38
rs761271060 Chr.1: 34197921-34197921 on Build GRCh38
rs758352765 Chr.1: 34197785-34197785 on Build GRCh38
rs763385816 Chr.1: 34197725-34197725 on Build GRCh38
rs1009406989 Chr.1: 34197723-34197723 on Build GRCh38
rs779214048 Chr.1: 34197895-34197895 on Build GRCh38
rs764819754 Chr.1: 34197855-34197855 on Build GRCh38
rs760290433 Chr.1: 34197817-34197817 on Build GRCh38
rs558422933 Chr.1: 34197910-34197910 on Build GRCh38

View Full Product Details