Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19851483-19851744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C22orf29
Entrez Gene ID
79680
Gene Name
chromosome 22 open reading frame 29
Gene Aliases
BOP
UniGene
Hs.105642
Gene Chromosome Location
Chr.22: 19846138-19854848 on Build GRCh38
Transcripts
NM_024627.5 NM_053004.2

Gene Symbol
GNB1L
Entrez Gene ID
54584
Gene Name
G protein subunit beta 1 like
Gene Aliases
DGCRK3, FKSG1, GY2, WDR14, WDVCF
UniGene
Hs.105642
Gene Chromosome Location
Chr.22: 19788411-19854939 on Build GRCh38
Transcripts
NM_024627.5 NM_053004.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
725597 c.661G>C p.E221Q Chr.22: 19851601-19851601 on Build GRCh38
4933288 c.747A>G p.V249V Chr.22: 19851515-19851515 on Build GRCh38
7281918 c.729G>A p.V243V Chr.22: 19851533-19851533 on Build GRCh38
6615672 c.615G>A p.V205V Chr.22: 19851647-19851647 on Build GRCh38
5499433 c.745_746delGT p.V249fs*2 Chr.22: 19851516-19851517 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1049784546 Chr.22: 19851660-19851660 on Build GRCh38
rs138140294 Chr.22: 19851713-19851713 on Build GRCh38
rs779045629 Chr.22: 19851509-19851509 on Build GRCh38
rs577981288 Chr.22: 19851553-19851553 on Build GRCh38
rs753038784 Chr.22: 19851596-19851596 on Build GRCh38
rs749223065 Chr.22: 19851631-19851631 on Build GRCh38
rs202242238 Chr.22: 19851590-19851590 on Build GRCh38
rs778085955 Chr.22: 19851603-19851603 on Build GRCh38
rs771723218 Chr.22: 19851670-19851670 on Build GRCh38
rs753902430 Chr.22: 19851579-19851579 on Build GRCh38
rs773906181 Chr.22: 19851640-19851640 on Build GRCh38
rs752842468 Chr.22: 19851717-19851717 on Build GRCh38
rs543070038 Chr.22: 19851554-19851554 on Build GRCh38
rs755930384 Chr.22: 19851617-19851617 on Build GRCh38
rs768105505 Chr.22: 19851639-19851639 on Build GRCh38
rs775451854 Chr.22: 19851546-19851546 on Build GRCh38
rs147446411 Chr.22: 19851517-19851517 on Build GRCh38
rs996356170 Chr.22: 19851657-19851657 on Build GRCh38
rs758702271 Chr.22: 19851601-19851601 on Build GRCh38
rs1005457816 Chr.22: 19851694-19851694 on Build GRCh38
rs754341627 Chr.22: 19851586-19851586 on Build GRCh38
rs781106314 Chr.22: 19851608-19851608 on Build GRCh38
rs772801297 Chr.22: 19851671-19851671 on Build GRCh38
rs777062618 Chr.22: 19851701-19851701 on Build GRCh38
rs762534530 Chr.22: 19851629-19851629 on Build GRCh38
rs747590171 Chr.22: 19851645-19851645 on Build GRCh38
rs765347164 Chr.22: 19851589-19851589 on Build GRCh38
rs61754879 Chr.22: 19851587-19851587 on Build GRCh38
rs773569158 Chr.22: 19851571-19851571 on Build GRCh38
rs771033342 Chr.22: 19851706-19851707 on Build GRCh38
rs576685823 Chr.22: 19851522-19851522 on Build GRCh38
rs760084173 Chr.22: 19851708-19851708 on Build GRCh38
rs766745486 Chr.22: 19851678-19851678 on Build GRCh38
rs760970479 Chr.22: 19851577-19851577 on Build GRCh38
rs944999348 Chr.22: 19851573-19851573 on Build GRCh38
rs944494033 Chr.22: 19851618-19851618 on Build GRCh38
rs747265411 Chr.22: 19851633-19851650 on Build GRCh38
rs766827661 Chr.22: 19851580-19851580 on Build GRCh38
rs745595271 Chr.22: 19851532-19851532 on Build GRCh38
rs779893435 Chr.22: 19851624-19851624 on Build GRCh38
rs781133599 Chr.22: 19851527-19851527 on Build GRCh38
rs1052835515 Chr.22: 19851585-19851585 on Build GRCh38
rs182295773 Chr.22: 19851718-19851718 on Build GRCh38
rs904301740 Chr.22: 19851706-19851706 on Build GRCh38
rs761068877 Chr.22: 19851677-19851677 on Build GRCh38
rs200035385 Chr.22: 19851549-19851549 on Build GRCh38
rs748380390 Chr.22: 19851511-19851511 on Build GRCh38
rs1021388803 Chr.22: 19851666-19851666 on Build GRCh38
rs767936404 Chr.22: 19851560-19851560 on Build GRCh38
rs769868190 Chr.22: 19851543-19851543 on Build GRCh38
rs932180181 Chr.22: 19851544-19851544 on Build GRCh38

View Full Product Details