Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 104891185-104891434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP12
Entrez Gene ID
100506742
Gene Name
caspase 12 (gene/pseudogene)
Gene Aliases
CASP-12, CASP12P1
UniGene
Hs.476989
Gene Chromosome Location
Chr.11: 104885718-104898670 on Build GRCh38
Transcripts
NM_001191016.2 NR_034061.2 NR_034063.2 NR_034064.2 NR_034065.2 NR_034066.2 NR_034067.2 NR_034068.2 NR_034070.2 NR_034071.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5890565 c.565G>A p.D189N Chr.11: 104891272-104891272 on Build GRCh38
6519532 c.499C>T p.L167L Chr.11: 104891338-104891338 on Build GRCh38
6574904 c.648C>T p.I216I Chr.11: 104891189-104891189 on Build GRCh38
2109041 c.482C>A p.S161Y Chr.11: 104891355-104891355 on Build GRCh38
6552622 c.498C>A p.F166L Chr.11: 104891339-104891339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751565046 Chr.11: 104891360-104891360 on Build GRCh38
rs776049923 Chr.11: 104891272-104891272 on Build GRCh38
rs766047352 Chr.11: 104891346-104891346 on Build GRCh38
rs976183650 Chr.11: 104891371-104891371 on Build GRCh38
rs189852923 Chr.11: 104891261-104891261 on Build GRCh38
rs199637202 Chr.11: 104891222-104891222 on Build GRCh38
rs866320801 Chr.11: 104891390-104891390 on Build GRCh38
rs764136136 Chr.11: 104891322-104891322 on Build GRCh38
rs764978278 Chr.11: 104891365-104891365 on Build GRCh38
rs887912779 Chr.11: 104891410-104891410 on Build GRCh38
rs1000818106 Chr.11: 104891303-104891303 on Build GRCh38
rs553678237 Chr.11: 104891214-104891214 on Build GRCh38
rs761518202 Chr.11: 104891281-104891281 on Build GRCh38
rs753753798 Chr.11: 104891323-104891323 on Build GRCh38
rs895592278 Chr.11: 104891233-104891233 on Build GRCh38
rs906040783 Chr.11: 104891278-104891278 on Build GRCh38
rs781427729 Chr.11: 104891377-104891377 on Build GRCh38
rs980868478 Chr.11: 104891301-104891301 on Build GRCh38
rs185140650 Chr.11: 104891230-104891230 on Build GRCh38
rs774893448 Chr.11: 104891245-104891245 on Build GRCh38
rs764927653 Chr.11: 104891287-104891287 on Build GRCh38
rs955487080 Chr.11: 104891374-104891374 on Build GRCh38
rs985515920 Chr.11: 104891381-104891381 on Build GRCh38
rs777257486 Chr.11: 104891229-104891229 on Build GRCh38
rs140309344 Chr.11: 104891260-104891260 on Build GRCh38
rs931710516 Chr.11: 104891379-104891379 on Build GRCh38
rs200223916 Chr.11: 104891335-104891335 on Build GRCh38
rs765014132 Chr.11: 104891259-104891260 on Build GRCh38
rs149162579 Chr.11: 104891370-104891370 on Build GRCh38
rs145290992 Chr.11: 104891342-104891342 on Build GRCh38
rs575832046 Chr.11: 104891283-104891283 on Build GRCh38
rs752822044 Chr.11: 104891385-104891385 on Build GRCh38
rs780338396 Chr.11: 104891238-104891238 on Build GRCh38
rs112756694 Chr.11: 104891345-104891345 on Build GRCh38

View Full Product Details