Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4890992-4891253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARRDC5
Entrez Gene ID
645432
Gene Name
arrestin domain containing 5
Gene Aliases
-
UniGene
Hs.574574
Gene Chromosome Location
Chr.19: 4867614-4915469 on Build GRCh38
Transcripts
NM_001080523.1 XM_005259616.2 XM_011528181.2 XM_017027144.1 XM_017027145.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3536490 c.856G>A p.G286S Chr.19: 4891219-4891219 on Build GRCh38
474983 c.968C>T p.S323F Chr.19: 4891107-4891107 on Build GRCh38
3536489 c.927G>A p.K309K Chr.19: 4891148-4891148 on Build GRCh38
3371400 c.1004C>T p.P335L Chr.19: 4891071-4891071 on Build GRCh38
4749107 c.1015G>A p.D339N Chr.19: 4891060-4891060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756973291 Chr.19: 4891144-4891144 on Build GRCh38
rs755694883 Chr.19: 4891062-4891062 on Build GRCh38
rs367940039 Chr.19: 4891097-4891097 on Build GRCh38
rs768795394 Chr.19: 4891102-4891102 on Build GRCh38
rs771529783 Chr.19: 4891040-4891040 on Build GRCh38
rs753592663 Chr.19: 4891070-4891070 on Build GRCh38
rs747577618 Chr.19: 4891167-4891167 on Build GRCh38
rs376606615 Chr.19: 4891236-4891236 on Build GRCh38
rs374918303 Chr.19: 4891082-4891082 on Build GRCh38
rs758453263 Chr.19: 4891133-4891134 on Build GRCh38
rs939194068 Chr.19: 4891200-4891200 on Build GRCh38
rs543694184 Chr.19: 4891052-4891052 on Build GRCh38
rs751292533 Chr.19: 4891039-4891039 on Build GRCh38
rs763754851 Chr.19: 4891131-4891131 on Build GRCh38
rs750143483 Chr.19: 4891201-4891201 on Build GRCh38
rs766783093 Chr.19: 4891204-4891204 on Build GRCh38
rs763226223 Chr.19: 4891025-4891025 on Build GRCh38
rs753263919 Chr.19: 4891139-4891139 on Build GRCh38
rs752884363 Chr.19: 4891052-4891052 on Build GRCh38
rs768442198 Chr.19: 4891229-4891229 on Build GRCh38
rs375360455 Chr.19: 4891180-4891180 on Build GRCh38
rs370201794 Chr.19: 4891069-4891069 on Build GRCh38
rs369148360 Chr.19: 4891207-4891207 on Build GRCh38
rs778012837 Chr.19: 4891227-4891228 on Build GRCh38
rs774736367 Chr.19: 4891081-4891081 on Build GRCh38
rs368428633 Chr.19: 4891164-4891164 on Build GRCh38
rs375874953 Chr.19: 4891206-4891206 on Build GRCh38
rs564754641 Chr.19: 4891181-4891181 on Build GRCh38
rs763905311 Chr.19: 4891186-4891186 on Build GRCh38
rs776563345 Chr.19: 4891105-4891105 on Build GRCh38
rs767590993 Chr.19: 4891042-4891042 on Build GRCh38
rs373336077 Chr.19: 4891226-4891226 on Build GRCh38
rs775411880 Chr.19: 4891184-4891184 on Build GRCh38
rs372032071 Chr.19: 4891170-4891170 on Build GRCh38
rs781568595 Chr.19: 4891211-4891211 on Build GRCh38
rs781375501 Chr.19: 4891171-4891171 on Build GRCh38
rs371210013 Chr.19: 4891203-4891203 on Build GRCh38
rs766257003 Chr.19: 4891019-4891019 on Build GRCh38
rs267605523 Chr.19: 4891163-4891163 on Build GRCh38
rs947787411 Chr.19: 4891190-4891190 on Build GRCh38
rs559756238 Chr.19: 4891193-4891193 on Build GRCh38
rs564002058 Chr.19: 4891120-4891120 on Build GRCh38
rs779382244 Chr.19: 4891076-4891076 on Build GRCh38
rs543324281 Chr.19: 4891182-4891182 on Build GRCh38
rs374124848 Chr.19: 4891158-4891158 on Build GRCh38
rs762147753 Chr.19: 4891115-4891115 on Build GRCh38
rs529524038 Chr.19: 4891166-4891166 on Build GRCh38
rs2297506 Chr.19: 4891227-4891227 on Build GRCh38
rs199933763 Chr.19: 4891195-4891195 on Build GRCh38
rs897720059 Chr.19: 4891212-4891212 on Build GRCh38
rs756845608 Chr.19: 4891074-4891074 on Build GRCh38
rs372125196 Chr.19: 4891121-4891121 on Build GRCh38
rs764738190 Chr.19: 4891196-4891196 on Build GRCh38
rs760685125 Chr.19: 4891125-4891125 on Build GRCh38
rs769878777 Chr.19: 4891119-4891119 on Build GRCh38
rs752682959 Chr.19: 4891123-4891123 on Build GRCh38
rs756148998 Chr.19: 4891214-4891214 on Build GRCh38
rs747450378 Chr.19: 4891086-4891086 on Build GRCh38
rs769861681 Chr.19: 4891177-4891177 on Build GRCh38
rs748205631 Chr.19: 4891175-4891175 on Build GRCh38
rs373562426 Chr.19: 4891118-4891118 on Build GRCh38
rs777901315 Chr.19: 4891220-4891220 on Build GRCh38

View Full Product Details