Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7743116-7743341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD209
Entrez Gene ID
30835
Gene Name
CD209 molecule
Gene Aliases
CDSIGN, CLEC4L, DC-SIGN, DC-SIGN1
UniGene
Hs.278694
Gene Chromosome Location
Chr.19: 7739993-7747609 on Build GRCh38
Transcripts
NM_001144893.1 NM_001144894.1 NM_001144895.1 NM_001144896.1 NM_001144897.1 NM_001144899.1 NM_021155.3 NR_026692.1 XM_005272472.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4857348 c.971C>T p.P324L Chr.19: 7743211-7743211 on Build GRCh38
5973251 c.1048T>C p.F350L Chr.19: 7743134-7743134 on Build GRCh38
127575 c.1095C>T p.N365N Chr.19: 7743159-7743159 on Build GRCh38
4897773 c.1000G>A p.E334K Chr.19: 7743182-7743182 on Build GRCh38
7170224 c.996C>T p.C332C Chr.19: 7743186-7743186 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767685744 Chr.19: 7743181-7743181 on Build GRCh38
rs772476580 Chr.19: 7743204-7743204 on Build GRCh38
rs149318064 Chr.19: 7743150-7743150 on Build GRCh38
rs756319987 Chr.19: 7743260-7743260 on Build GRCh38
rs554926524 Chr.19: 7743311-7743311 on Build GRCh38
rs531513041 Chr.19: 7743272-7743273 on Build GRCh38
rs947025444 Chr.19: 7743158-7743158 on Build GRCh38
rs935520212 Chr.19: 7743226-7743226 on Build GRCh38
rs868066140 Chr.19: 7743242-7743242 on Build GRCh38
rs779286111 Chr.19: 7743269-7743269 on Build GRCh38
rs774079703 Chr.19: 7743196-7743196 on Build GRCh38
rs532110467 Chr.19: 7743233-7743233 on Build GRCh38
rs200282091 Chr.19: 7743185-7743185 on Build GRCh38
rs77505063 Chr.19: 7743278-7743279 on Build GRCh38
rs761614211 Chr.19: 7743157-7743157 on Build GRCh38
rs749723284 Chr.19: 7743145-7743145 on Build GRCh38
rs867029884 Chr.19: 7743220-7743220 on Build GRCh38
rs372037869 Chr.19: 7743275-7743275 on Build GRCh38
rs553008652 Chr.19: 7743156-7743156 on Build GRCh38
rs568970102 Chr.19: 7743268-7743268 on Build GRCh38
rs754983304 Chr.19: 7743175-7743175 on Build GRCh38
rs765057843 Chr.19: 7743248-7743248 on Build GRCh38
rs750442689 Chr.19: 7743174-7743174 on Build GRCh38
rs901004109 Chr.19: 7743223-7743223 on Build GRCh38
rs752762064 Chr.19: 7743258-7743258 on Build GRCh38
rs897188297 Chr.19: 7743302-7743302 on Build GRCh38
rs368570264 Chr.19: 7743184-7743184 on Build GRCh38
rs773003473 Chr.19: 7743251-7743251 on Build GRCh38
rs201898991 Chr.19: 7743218-7743218 on Build GRCh38
rs779527826 Chr.19: 7743198-7743198 on Build GRCh38
rs776327212 Chr.19: 7743227-7743227 on Build GRCh38
rs375340194 Chr.19: 7743287-7743287 on Build GRCh38
rs749655694 Chr.19: 7743186-7743186 on Build GRCh38
rs368971770 Chr.19: 7743256-7743256 on Build GRCh38
rs747849049 Chr.19: 7743285-7743285 on Build GRCh38
rs1049550237 Chr.19: 7743252-7743252 on Build GRCh38
rs754150508 Chr.19: 7743266-7743266 on Build GRCh38
rs779057361 Chr.19: 7743292-7743293 on Build GRCh38
rs764405668 Chr.19: 7743263-7743263 on Build GRCh38
rs376393724 Chr.19: 7743255-7743255 on Build GRCh38
rs746434437 Chr.19: 7743202-7743202 on Build GRCh38
rs780441267 Chr.19: 7743276-7743276 on Build GRCh38
rs746508965 Chr.19: 7743217-7743217 on Build GRCh38
rs765147780 Chr.19: 7743166-7743166 on Build GRCh38
rs775028274 Chr.19: 7743215-7743215 on Build GRCh38
rs769459892 Chr.19: 7743286-7743286 on Build GRCh38
rs35545365 Chr.19: 7743183-7743183 on Build GRCh38
rs761593739 Chr.19: 7743243-7743243 on Build GRCh38
rs774981477 Chr.19: 7743147-7743147 on Build GRCh38
rs11465392 Chr.19: 7743273-7743273 on Build GRCh38
rs771648411 Chr.19: 7743143-7743143 on Build GRCh38
rs945126737 Chr.19: 7743203-7743203 on Build GRCh38
rs11465391 Chr.19: 7743277-7743277 on Build GRCh38
rs763770706 Chr.19: 7743155-7743155 on Build GRCh38
rs779368115 Chr.19: 7743304-7743304 on Build GRCh38

View Full Product Details