Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81103830-81104067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAIAP2
Entrez Gene ID
10458
Gene Name
BAI1 associated protein 2
Gene Aliases
BAP2, FLAF3, IRSP53
UniGene
Hs.128316
Gene Chromosome Location
Chr.17: 81035147-81117432 on Build GRCh38
Transcripts
NM_001144888.1 NM_006340.2 NM_017450.2 NM_017451.2 XM_005256943.1 XM_005256944.1 XM_005256945.1 XM_005256948.3 XM_006721635.1 XM_006721636.1 XM_006721637.2 XM_011524193.1 XM_011524194.1 XM_011524195.1 XM_011524196.1 XM_011524197.1 XM_017024017.1 XM_017024018.1 XM_017024019.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2804401 c.992G>A p.S331N Chr.17: 81104034-81104034 on Build GRCh38
1750402 c.1011C>T p.Y337Y Chr.17: 81104053-81104053 on Build GRCh38
5594255 c.968C>T p.S323F Chr.17: 81104010-81104010 on Build GRCh38
7288691 c.969C>T p.S323S Chr.17: 81104011-81104011 on Build GRCh38
2804395 c.948C>T p.R316R Chr.17: 81103990-81103990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767893587 Chr.17: 81103883-81103883 on Build GRCh38
rs377680789 Chr.17: 81103975-81103975 on Build GRCh38
rs577903416 Chr.17: 81103913-81103913 on Build GRCh38
rs746723636 Chr.17: 81103928-81103928 on Build GRCh38
rs547208989 Chr.17: 81104038-81104038 on Build GRCh38
rs951245978 Chr.17: 81103867-81103867 on Build GRCh38
rs758423099 Chr.17: 81103966-81103966 on Build GRCh38
rs112574740 Chr.17: 81103863-81103863 on Build GRCh38
rs944066804 Chr.17: 81103952-81103952 on Build GRCh38
rs761516246 Chr.17: 81103852-81103852 on Build GRCh38
rs752473401 Chr.17: 81104044-81104044 on Build GRCh38
rs142250650 Chr.17: 81103957-81103957 on Build GRCh38
rs781113973 Chr.17: 81103859-81103859 on Build GRCh38
rs561013874 Chr.17: 81104013-81104013 on Build GRCh38
rs757348479 Chr.17: 81103899-81103899 on Build GRCh38
rs775928425 Chr.17: 81104008-81104008 on Build GRCh38
rs764796647 Chr.17: 81103866-81103866 on Build GRCh38
rs780198712 Chr.17: 81103972-81103972 on Build GRCh38
rs141436998 Chr.17: 81103946-81103946 on Build GRCh38
rs762128990 Chr.17: 81104017-81104017 on Build GRCh38
rs748742558 Chr.17: 81104004-81104004 on Build GRCh38
rs776209043 Chr.17: 81103931-81103931 on Build GRCh38
rs772657995 Chr.17: 81103942-81103942 on Build GRCh38
rs148408272 Chr.17: 81104023-81104023 on Build GRCh38
rs1023609159 Chr.17: 81104045-81104045 on Build GRCh38
rs781015961 Chr.17: 81103978-81103978 on Build GRCh38
rs753023776 Chr.17: 81103884-81103884 on Build GRCh38
rs768126090 Chr.17: 81103862-81103862 on Build GRCh38
rs754765663 Chr.17: 81103977-81103977 on Build GRCh38
rs528172800 Chr.17: 81104022-81104022 on Build GRCh38
rs756367448 Chr.17: 81103886-81103886 on Build GRCh38
rs751490909 Chr.17: 81103956-81103956 on Build GRCh38
rs768607123 Chr.17: 81103908-81103908 on Build GRCh38
rs761006511 Chr.17: 81103949-81103949 on Build GRCh38
rs371249844 Chr.17: 81104016-81104016 on Build GRCh38
rs747775919 Chr.17: 81103989-81103989 on Build GRCh38
rs369353973 Chr.17: 81103874-81103874 on Build GRCh38
rs751497094 Chr.17: 81104033-81104033 on Build GRCh38
rs935500919 Chr.17: 81103963-81103963 on Build GRCh38
rs912283617 Chr.17: 81103877-81103877 on Build GRCh38
rs1041492778 Chr.17: 81103955-81103955 on Build GRCh38
rs540871091 Chr.17: 81103859-81103859 on Build GRCh38
rs373571526 Chr.17: 81103882-81103882 on Build GRCh38
rs772717162 Chr.17: 81103992-81103992 on Build GRCh38
rs760433984 Chr.17: 81103861-81103861 on Build GRCh38
rs757535912 Chr.17: 81103937-81103937 on Build GRCh38
rs752448517 Chr.17: 81103859-81103860 on Build GRCh38
rs754048103 Chr.17: 81103953-81103953 on Build GRCh38
rs146135495 Chr.17: 81103945-81103945 on Build GRCh38
rs779998520 Chr.17: 81103926-81103926 on Build GRCh38
rs768483453 Chr.17: 81103930-81103930 on Build GRCh38
rs775849748 Chr.17: 81103948-81103948 on Build GRCh38
rs770418837 Chr.17: 81104005-81104005 on Build GRCh38
rs754078158 Chr.17: 81103896-81103896 on Build GRCh38
rs189890121 Chr.17: 81103887-81103887 on Build GRCh38
rs761214201 Chr.17: 81104011-81104011 on Build GRCh38
rs142524547 Chr.17: 81104034-81104034 on Build GRCh38
rs752098024 Chr.17: 81103873-81103873 on Build GRCh38
rs750460687 Chr.17: 81103964-81103964 on Build GRCh38
rs544940030 Chr.17: 81103936-81103936 on Build GRCh38
rs745852347 Chr.17: 81103911-81103911 on Build GRCh38
rs746958217 Chr.17: 81103976-81103976 on Build GRCh38
rs761284106 Chr.17: 81103864-81103864 on Build GRCh38
rs201778887 Chr.17: 81104043-81104043 on Build GRCh38
rs764516093 Chr.17: 81103951-81103951 on Build GRCh38
rs866018129 Chr.17: 81103994-81103994 on Build GRCh38

View Full Product Details