Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 151738359-151738616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGA2
Entrez Gene ID
1260
Gene Name
cyclic nucleotide gated channel alpha 2
Gene Aliases
CNCA, CNCA1, CNG2, OCNC1, OCNCALPHA, OCNCa
UniGene
Hs.447360
Gene Chromosome Location
Chr.X: 151734746-151745564 on Build GRCh38
Transcripts
NM_005140.1

SNP Detail

SNP ID Chromosome Location
rs952741761 Chr.X: 151738531-151738531 on Build GRCh38
rs1033589039 Chr.X: 151738490-151738491 on Build GRCh38
rs746796108 Chr.X: 151738506-151738506 on Build GRCh38
rs757386545 Chr.X: 151738436-151738436 on Build GRCh38
rs906477852 Chr.X: 151738484-151738484 on Build GRCh38
rs146494392 Chr.X: 151738587-151738587 on Build GRCh38
rs754263915 Chr.X: 151738564-151738564 on Build GRCh38
rs139232978 Chr.X: 151738493-151738493 on Build GRCh38
rs774269466 Chr.X: 151738516-151738516 on Build GRCh38
rs761937144 Chr.X: 151738517-151738517 on Build GRCh38
rs755412260 Chr.X: 151738565-151738565 on Build GRCh38
rs747735929 Chr.X: 151738496-151738496 on Build GRCh38
rs777407062 Chr.X: 151738400-151738400 on Build GRCh38
rs773056408 Chr.X: 151738505-151738505 on Build GRCh38
rs750769746 Chr.X: 151738440-151738440 on Build GRCh38
rs751965597 Chr.X: 151738512-151738512 on Build GRCh38
rs780405357 Chr.X: 151738470-151738470 on Build GRCh38
rs779524501 Chr.X: 151738583-151738583 on Build GRCh38
rs771830593 Chr.X: 151738497-151738497 on Build GRCh38
rs886081849 Chr.X: 151738419-151738419 on Build GRCh38
rs765259218 Chr.X: 151738514-151738514 on Build GRCh38
rs965354592 Chr.X: 151738552-151738552 on Build GRCh38
rs150994680 Chr.X: 151738544-151738544 on Build GRCh38
rs188269270 Chr.X: 151738528-151738528 on Build GRCh38
rs1042007541 Chr.X: 151738382-151738382 on Build GRCh38
rs1018596411 Chr.X: 151738501-151738501 on Build GRCh38
rs148854921 Chr.X: 151738404-151738404 on Build GRCh38
rs764000814 Chr.X: 151738435-151738435 on Build GRCh38
rs142562634 Chr.X: 151738527-151738527 on Build GRCh38
rs200529056 Chr.X: 151738589-151738589 on Build GRCh38
rs201028264 Chr.X: 151738455-151738455 on Build GRCh38
rs762655246 Chr.X: 151738414-151738414 on Build GRCh38
rs1028778052 Chr.X: 151738535-151738535 on Build GRCh38
rs769231187 Chr.X: 151738490-151738490 on Build GRCh38
rs766685052 Chr.X: 151738563-151738563 on Build GRCh38
rs758048775 Chr.X: 151738588-151738588 on Build GRCh38
rs770792204 Chr.X: 151738507-151738507 on Build GRCh38
rs145435266 Chr.X: 151738489-151738489 on Build GRCh38
rs781439227 Chr.X: 151738438-151738438 on Build GRCh38

View Full Product Details