Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 90268219-90268474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP69
Entrez Gene ID
79846
Gene Name
cilia and flagella associated protein 69
Gene Aliases
C7orf63, FAP69
UniGene
Hs.521014
Gene Chromosome Location
Chr.7: 90245174-90320964 on Build GRCh38
Transcripts
NM_001039706.2 NM_001160138.1 XM_005250600.3 XM_011516577.2 XM_011516578.2 XM_011516579.1 XM_017012624.1 XM_017012625.1 XM_017012626.1 XM_017012627.1 XM_017012628.1 XM_017012629.1 XM_017012630.1 XM_017012631.1 XM_017012632.1 XM_017012633.1 XM_017012634.1 XM_017012635.1 XM_017012636.1 XM_017012637.1 XM_017012638.1 XM_017012639.1 XM_017012641.1 XM_017012642.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3721089 c.467G>T p.R156M Chr.7: 90268319-90268319 on Build GRCh38
1635463 c.512C>T p.P171L Chr.7: 90268364-90268364 on Build GRCh38
3642129 c.493G>T p.D165Y Chr.7: 90268345-90268345 on Build GRCh38
5408229 c.439T>C p.L147L Chr.7: 90268291-90268291 on Build GRCh38
7111823 c.444G>A p.M148I Chr.7: 90268296-90268296 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs574843478 Chr.7: 90268443-90268443 on Build GRCh38
rs778034979 Chr.7: 90268402-90268402 on Build GRCh38
rs1010527727 Chr.7: 90268439-90268439 on Build GRCh38
rs368604033 Chr.7: 90268395-90268395 on Build GRCh38
rs914202498 Chr.7: 90268441-90268441 on Build GRCh38
rs971055429 Chr.7: 90268444-90268444 on Build GRCh38
rs1054222402 Chr.7: 90268440-90268440 on Build GRCh38
rs745864940 Chr.7: 90268283-90268283 on Build GRCh38
rs746843204 Chr.7: 90268301-90268301 on Build GRCh38
rs1048175982 Chr.7: 90268381-90268381 on Build GRCh38
rs777984596 Chr.7: 90268282-90268282 on Build GRCh38
rs979882681 Chr.7: 90268322-90268322 on Build GRCh38
rs772680500 Chr.7: 90268349-90268349 on Build GRCh38
rs755378721 Chr.7: 90268362-90268362 on Build GRCh38
rs776348740 Chr.7: 90268342-90268342 on Build GRCh38
rs778795185 Chr.7: 90268317-90268317 on Build GRCh38
rs767718862 Chr.7: 90268379-90268379 on Build GRCh38
rs1016650925 Chr.7: 90268269-90268269 on Build GRCh38
rs751974341 Chr.7: 90268356-90268356 on Build GRCh38
rs748254646 Chr.7: 90268255-90268255 on Build GRCh38
rs368893731 Chr.7: 90268266-90268266 on Build GRCh38
rs749353784 Chr.7: 90268411-90268411 on Build GRCh38
rs371422121 Chr.7: 90268431-90268431 on Build GRCh38
rs781512331 Chr.7: 90268252-90268252 on Build GRCh38
rs17863057 Chr.7: 90268389-90268389 on Build GRCh38
rs371357343 Chr.7: 90268434-90268434 on Build GRCh38
rs376199817 Chr.7: 90268344-90268344 on Build GRCh38
rs373747438 Chr.7: 90268425-90268425 on Build GRCh38
rs200834034 Chr.7: 90268352-90268352 on Build GRCh38
rs1039053774 Chr.7: 90268427-90268427 on Build GRCh38
rs201631766 Chr.7: 90268354-90268354 on Build GRCh38
rs780170968 Chr.7: 90268422-90268422 on Build GRCh38
rs772100468 Chr.7: 90268288-90268288 on Build GRCh38
rs372422328 Chr.7: 90268315-90268315 on Build GRCh38
rs1033168556 Chr.7: 90268436-90268436 on Build GRCh38
rs764795583 Chr.7: 90268346-90268346 on Build GRCh38
rs17869450 Chr.7: 90268437-90268437 on Build GRCh38
rs927039317 Chr.7: 90268323-90268323 on Build GRCh38
rs530113668 Chr.7: 90268331-90268331 on Build GRCh38
rs775471186 Chr.7: 90268295-90268295 on Build GRCh38
rs17867345 Chr.7: 90268409-90268409 on Build GRCh38
rs757363115 Chr.7: 90268415-90268415 on Build GRCh38
rs376739951 Chr.7: 90268426-90268426 on Build GRCh38

View Full Product Details