Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 81046184-81046445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP3
Entrez Gene ID
651
Gene Name
bone morphogenetic protein 3
Gene Aliases
BMP-3A
UniGene
Hs.387411
Gene Chromosome Location
Chr.4: 81030778-81057531 on Build GRCh38
Transcripts
NM_001201.2 XM_006714291.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6611944 c.886C>A p.P296T Chr.4: 81046307-81046307 on Build GRCh38
4665839 c.910G>T p.G304W Chr.4: 81046331-81046331 on Build GRCh38
4126246 c.839C>T p.A280V Chr.4: 81046260-81046260 on Build GRCh38
4126247 c.938A>C p.E313A Chr.4: 81046359-81046359 on Build GRCh38
5975224 c.854G>A p.R285Q Chr.4: 81046275-81046275 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs79405615 Chr.4: 81046238-81046238 on Build GRCh38
rs755609561 Chr.4: 81046370-81046370 on Build GRCh38
rs779660778 Chr.4: 81046286-81046286 on Build GRCh38
rs776862828 Chr.4: 81046241-81046241 on Build GRCh38
rs763896031 Chr.4: 81046343-81046343 on Build GRCh38
rs759780251 Chr.4: 81046252-81046252 on Build GRCh38
rs751449569 Chr.4: 81046272-81046272 on Build GRCh38
rs763092557 Chr.4: 81046340-81046340 on Build GRCh38
rs1017463616 Chr.4: 81046379-81046379 on Build GRCh38
rs370097482 Chr.4: 81046367-81046367 on Build GRCh38
rs578226153 Chr.4: 81046401-81046401 on Build GRCh38
rs765453612 Chr.4: 81046333-81046333 on Build GRCh38
rs775853500 Chr.4: 81046334-81046334 on Build GRCh38
rs767410889 Chr.4: 81046353-81046353 on Build GRCh38
rs771363638 Chr.4: 81046325-81046325 on Build GRCh38
rs200302858 Chr.4: 81046208-81046208 on Build GRCh38
rs17005033 Chr.4: 81046285-81046285 on Build GRCh38
rs753005915 Chr.4: 81046261-81046261 on Build GRCh38
rs773754745 Chr.4: 81046222-81046222 on Build GRCh38
rs773451656 Chr.4: 81046309-81046309 on Build GRCh38
rs541364851 Chr.4: 81046346-81046346 on Build GRCh38
rs150626762 Chr.4: 81046388-81046388 on Build GRCh38
rs759652850 Chr.4: 81046329-81046329 on Build GRCh38
rs758894719 Chr.4: 81046243-81046243 on Build GRCh38
rs747491518 Chr.4: 81046320-81046320 on Build GRCh38
rs975961336 Chr.4: 81046209-81046209 on Build GRCh38
rs368791166 Chr.4: 81046275-81046275 on Build GRCh38
rs758186711 Chr.4: 81046262-81046262 on Build GRCh38
rs766414400 Chr.4: 81046237-81046237 on Build GRCh38
rs866813886 Chr.4: 81046217-81046217 on Build GRCh38
rs372785256 Chr.4: 81046391-81046391 on Build GRCh38
rs781758330 Chr.4: 81046406-81046406 on Build GRCh38
rs61729827 Chr.4: 81046378-81046378 on Build GRCh38
rs772479539 Chr.4: 81046290-81046290 on Build GRCh38
rs143389795 Chr.4: 81046230-81046230 on Build GRCh38
rs950312043 Chr.4: 81046296-81046296 on Build GRCh38
rs1021330543 Chr.4: 81046405-81046405 on Build GRCh38
rs769931039 Chr.4: 81046414-81046414 on Build GRCh38
rs753419201 Chr.4: 81046385-81046385 on Build GRCh38
rs746432172 Chr.4: 81046409-81046409 on Build GRCh38
rs967821582 Chr.4: 81046232-81046232 on Build GRCh38
rs771362322 Chr.4: 81046404-81046404 on Build GRCh38
rs749000111 Chr.4: 81046289-81046289 on Build GRCh38
rs1048692199 Chr.4: 81046335-81046335 on Build GRCh38
rs140653938 Chr.4: 81046274-81046274 on Build GRCh38
rs372429165 Chr.4: 81046284-81046284 on Build GRCh38
rs765583405 Chr.4: 81046260-81046260 on Build GRCh38
rs138982746 Chr.4: 81046269-81046269 on Build GRCh38
rs188449581 Chr.4: 81046345-81046345 on Build GRCh38

View Full Product Details