Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184260188-184260461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMK2N2
Entrez Gene ID
94032
Gene Name
calcium/calmodulin dependent protein kinase II inhibitor 2
Gene Aliases
CAM-KIIN, CAMKIIN
UniGene
Hs.585003
Gene Chromosome Location
Chr.3: 184259215-184261463 on Build GRCh38
Transcripts
NM_033259.2 NM_014693.3

Gene Symbol
ECE2
Entrez Gene ID
9718
Gene Name
endothelin converting enzyme 2
Gene Aliases
-
UniGene
Hs.146161
Gene Chromosome Location
Chr.3: 184249657-184293031 on Build GRCh38
Transcripts
NM_033259.2 NM_014693.3

SNP Detail

SNP ID Chromosome Location
rs942683928 Chr.3: 184260393-184260393 on Build GRCh38
rs1002941412 Chr.3: 184260323-184260323 on Build GRCh38
rs974261510 Chr.3: 184260321-184260321 on Build GRCh38
rs764054440 Chr.3: 184260265-184260265 on Build GRCh38
rs947465253 Chr.3: 184260439-184260439 on Build GRCh38
rs1015998166 Chr.3: 184260259-184260259 on Build GRCh38
rs573653970 Chr.3: 184260418-184260418 on Build GRCh38
rs766164887 Chr.3: 184260245-184260245 on Build GRCh38
rs372889845 Chr.3: 184260289-184260289 on Build GRCh38
rs190455930 Chr.3: 184260369-184260369 on Build GRCh38
rs755000365 Chr.3: 184260263-184260263 on Build GRCh38
rs922990869 Chr.3: 184260434-184260434 on Build GRCh38
rs546698926 Chr.3: 184260305-184260305 on Build GRCh38
rs1035755698 Chr.3: 184260336-184260336 on Build GRCh38
rs569406055 Chr.3: 184260264-184260264 on Build GRCh38
rs764831945 Chr.3: 184260212-184260212 on Build GRCh38
rs555086351 Chr.3: 184260415-184260415 on Build GRCh38
rs1030772655 Chr.3: 184260373-184260373 on Build GRCh38
rs979786927 Chr.3: 184260370-184260370 on Build GRCh38
rs1012878414 Chr.3: 184260342-184260342 on Build GRCh38
rs762676759 Chr.3: 184260229-184260229 on Build GRCh38
rs1022081330 Chr.3: 184260365-184260365 on Build GRCh38
rs577485885 Chr.3: 184260220-184260220 on Build GRCh38
rs898026992 Chr.3: 184260295-184260295 on Build GRCh38
rs182094851 Chr.3: 184260395-184260395 on Build GRCh38
rs772874827 Chr.3: 184260219-184260219 on Build GRCh38
rs931726775 Chr.3: 184260437-184260437 on Build GRCh38
rs373116496 Chr.3: 184260337-184260337 on Build GRCh38
rs914653191 Chr.3: 184260397-184260397 on Build GRCh38

View Full Product Details