Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 309924-310181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIP2C
Entrez Gene ID
22982
Gene Name
disco interacting protein 2 homolog C
Gene Aliases
KIAA0934
UniGene
Hs.432397
Gene Chromosome Location
Chr.10: 274190-689710 on Build GRCh38
Transcripts
NM_014974.2 XM_005252426.3 XM_005252427.3 XM_005252428.4 XM_005252430.2 XM_011519428.2 XM_011519429.2 XM_011519430.2 XM_011519431.2 XM_011519432.1 XM_011519433.2 XM_017015977.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7086847 c.3935G>A p.G1312E Chr.10: 310082-310082 on Build GRCh38
917895 c.3981C>T p.H1327H Chr.10: 310036-310036 on Build GRCh38
1956028 c.3965T>C p.M1322T Chr.10: 310052-310052 on Build GRCh38
1956027 c.3986G>A p.R1329K Chr.10: 310031-310031 on Build GRCh38
5573374 c.3986+2T>C p.? Chr.10: 310029-310029 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866034398 Chr.10: 310055-310055 on Build GRCh38
rs184524196 Chr.10: 310132-310132 on Build GRCh38
rs573499024 Chr.10: 310150-310150 on Build GRCh38
rs776938070 Chr.10: 310045-310045 on Build GRCh38
rs748877695 Chr.10: 310007-310007 on Build GRCh38
rs751923146 Chr.10: 310118-310118 on Build GRCh38
rs45608133 Chr.10: 310140-310140 on Build GRCh38
rs191755936 Chr.10: 310111-310111 on Build GRCh38
rs897221870 Chr.10: 309955-309955 on Build GRCh38
rs187332353 Chr.10: 309999-309999 on Build GRCh38
rs768616009 Chr.10: 310035-310035 on Build GRCh38
rs754938192 Chr.10: 310141-310141 on Build GRCh38
rs779681538 Chr.10: 309990-309990 on Build GRCh38
rs369421614 Chr.10: 309969-309969 on Build GRCh38
rs374162032 Chr.10: 309990-309990 on Build GRCh38
rs776165992 Chr.10: 310066-310066 on Build GRCh38
rs777955587 Chr.10: 309982-309982 on Build GRCh38
rs138147106 Chr.10: 310134-310134 on Build GRCh38
rs759695360 Chr.10: 310059-310059 on Build GRCh38
rs763423347 Chr.10: 310105-310105 on Build GRCh38
rs150410788 Chr.10: 309977-309977 on Build GRCh38
rs200561296 Chr.10: 310036-310036 on Build GRCh38
rs769224552 Chr.10: 310003-310003 on Build GRCh38
rs761664186 Chr.10: 310038-310038 on Build GRCh38
rs528126847 Chr.10: 310120-310120 on Build GRCh38
rs757787462 Chr.10: 310060-310060 on Build GRCh38
rs753609319 Chr.10: 309978-309978 on Build GRCh38
rs778896889 Chr.10: 310143-310143 on Build GRCh38
rs751934482 Chr.10: 309985-309985 on Build GRCh38
rs750772904 Chr.10: 309982-309986 on Build GRCh38
rs757581603 Chr.10: 310122-310122 on Build GRCh38
rs749173027 Chr.10: 310017-310017 on Build GRCh38
rs891296602 Chr.10: 310123-310123 on Build GRCh38
rs771116499 Chr.10: 310041-310041 on Build GRCh38
rs780880911 Chr.10: 309993-309993 on Build GRCh38
rs756427621 Chr.10: 309989-309990 on Build GRCh38
rs200666436 Chr.10: 310016-310016 on Build GRCh38
rs377632100 Chr.10: 310108-310108 on Build GRCh38
rs767159040 Chr.10: 310129-310129 on Build GRCh38
rs112535917 Chr.10: 310024-310024 on Build GRCh38
rs34534378 Chr.10: 309981-309982 on Build GRCh38

View Full Product Details