Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 114597515-114597784 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1G1
Entrez Gene ID
9550
Gene Name
ATPase H+ transporting V1 subunit G1
Gene Aliases
ATP6G, ATP6G1, ATP6GL, ATP6J, Vma10
UniGene
Hs.388654
Gene Chromosome Location
Chr.9: 114587714-114598872 on Build GRCh38
Transcripts
NM_004888.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4664490 c.257C>T p.T86I Chr.9: 114597643-114597643 on Build GRCh38
5738707 c.344G>A p.R115H Chr.9: 114597730-114597730 on Build GRCh38
2151386 c.320G>A p.R107Q Chr.9: 114597706-114597706 on Build GRCh38
1104424 c.267G>A p.R89R Chr.9: 114597653-114597653 on Build GRCh38
223361 c.325G>A p.E109K Chr.9: 114597711-114597711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61742531 Chr.9: 114597652-114597652 on Build GRCh38
rs150320956 Chr.9: 114597738-114597738 on Build GRCh38
rs773730024 Chr.9: 114597547-114597547 on Build GRCh38
rs754577497 Chr.9: 114597729-114597729 on Build GRCh38
rs749641296 Chr.9: 114597744-114597745 on Build GRCh38
rs540425306 Chr.9: 114597705-114597705 on Build GRCh38
rs141395465 Chr.9: 114597641-114597641 on Build GRCh38
rs749551716 Chr.9: 114597736-114597736 on Build GRCh38
rs777982171 Chr.9: 114597638-114597638 on Build GRCh38
rs574916000 Chr.9: 114597698-114597698 on Build GRCh38
rs141529529 Chr.9: 114597743-114597743 on Build GRCh38
rs560343317 Chr.9: 114597716-114597716 on Build GRCh38
rs763968695 Chr.9: 114597594-114597594 on Build GRCh38
rs766532972 Chr.9: 114597561-114597561 on Build GRCh38
rs759628317 Chr.9: 114597650-114597650 on Build GRCh38
rs556510080 Chr.9: 114597611-114597611 on Build GRCh38
rs774285495 Chr.9: 114597565-114597565 on Build GRCh38
rs1037529851 Chr.9: 114597563-114597563 on Build GRCh38
rs570734987 Chr.9: 114597582-114597582 on Build GRCh38
rs201363023 Chr.9: 114597690-114597690 on Build GRCh38
rs778543044 Chr.9: 114597604-114597604 on Build GRCh38
rs760947184 Chr.9: 114597662-114597662 on Build GRCh38
rs756598486 Chr.9: 114597622-114597624 on Build GRCh38
rs753854212 Chr.9: 114597599-114597599 on Build GRCh38
rs757864164 Chr.9: 114597702-114597702 on Build GRCh38
rs1025449499 Chr.9: 114597651-114597651 on Build GRCh38
rs770315807 Chr.9: 114597624-114597624 on Build GRCh38
rs780759538 Chr.9: 114597637-114597637 on Build GRCh38
rs771698288 Chr.9: 114597542-114597542 on Build GRCh38
rs774490624 Chr.9: 114597644-114597644 on Build GRCh38
rs760493446 Chr.9: 114597591-114597591 on Build GRCh38
rs781771465 Chr.9: 114597617-114597617 on Build GRCh38
rs758062356 Chr.9: 114597614-114597614 on Build GRCh38
rs112425042 Chr.9: 114597635-114597635 on Build GRCh38
rs763341718 Chr.9: 114597556-114597556 on Build GRCh38
rs372130727 Chr.9: 114597546-114597546 on Build GRCh38
rs775810655 Chr.9: 114597656-114597656 on Build GRCh38
rs757117779 Chr.9: 114597603-114597603 on Build GRCh38
rs11539658 Chr.9: 114597633-114597633 on Build GRCh38
rs752550197 Chr.9: 114597590-114597590 on Build GRCh38
rs533936503 Chr.9: 114597543-114597543 on Build GRCh38
rs759906767 Chr.9: 114597573-114597573 on Build GRCh38
rs1013692981 Chr.9: 114597636-114597636 on Build GRCh38
rs745973549 Chr.9: 114597745-114597745 on Build GRCh38
rs765027930 Chr.9: 114597693-114597693 on Build GRCh38
rs773744869 Chr.9: 114597757-114597757 on Build GRCh38
rs369961991 Chr.9: 114597730-114597730 on Build GRCh38
rs11539656 Chr.9: 114597697-114597697 on Build GRCh38
rs768755489 Chr.9: 114597759-114597759 on Build GRCh38
rs746406910 Chr.9: 114597622-114597622 on Build GRCh38
rs538241798 Chr.9: 114597655-114597655 on Build GRCh38
rs199542651 Chr.9: 114597689-114597689 on Build GRCh38
rs745568729 Chr.9: 114597540-114597540 on Build GRCh38
rs772317365 Chr.9: 114597753-114597753 on Build GRCh38
rs539776456 Chr.9: 114597583-114597583 on Build GRCh38
rs145627791 Chr.9: 114597619-114597619 on Build GRCh38
rs921606307 Chr.9: 114597732-114597732 on Build GRCh38
rs139067587 Chr.9: 114597680-114597680 on Build GRCh38

View Full Product Details