Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 3638005-3638268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRACR2A
Entrez Gene ID
84766
Gene Name
calcium release activated channel regulator 2A
Gene Aliases
EFCAB4B
UniGene
Hs.504534
Gene Chromosome Location
Chr.12: 3613366-3753216 on Build GRCh38
Transcripts
NM_001144958.1 XM_006719021.3 XM_011521034.2 XM_011521036.2 XM_011521037.2 XM_011521038.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1586452 c.1573C>A p.Q525K Chr.12: 3638153-3638153 on Build GRCh38
7304684 c.1591G>A p.A531T Chr.12: 3638135-3638135 on Build GRCh38
4469305 c.1593C>T p.A531A Chr.12: 3638133-3638133 on Build GRCh38
3739618 c.1573C>T p.Q525* Chr.12: 3638153-3638153 on Build GRCh38
4468394 c.1541C>T p.P514L Chr.12: 3638185-3638185 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962413675 Chr.12: 3638213-3638213 on Build GRCh38
rs374954232 Chr.12: 3638194-3638194 on Build GRCh38
rs1010012564 Chr.12: 3638078-3638078 on Build GRCh38
rs767588976 Chr.12: 3638156-3638156 on Build GRCh38
rs193073745 Chr.12: 3638095-3638095 on Build GRCh38
rs748957811 Chr.12: 3638197-3638197 on Build GRCh38
rs923283391 Chr.12: 3638050-3638050 on Build GRCh38
rs755016204 Chr.12: 3638122-3638122 on Build GRCh38
rs760446541 Chr.12: 3638116-3638116 on Build GRCh38
rs935115227 Chr.12: 3638154-3638154 on Build GRCh38
rs780326925 Chr.12: 3638227-3638227 on Build GRCh38
rs1003497681 Chr.12: 3638114-3638114 on Build GRCh38
rs911152957 Chr.12: 3638206-3638206 on Build GRCh38
rs910840609 Chr.12: 3638069-3638069 on Build GRCh38
rs955235467 Chr.12: 3638190-3638190 on Build GRCh38
rs969498978 Chr.12: 3638158-3638159 on Build GRCh38
rs532414144 Chr.12: 3638158-3638158 on Build GRCh38
rs544454582 Chr.12: 3638188-3638188 on Build GRCh38
rs552177056 Chr.12: 3638226-3638226 on Build GRCh38
rs765432380 Chr.12: 3638084-3638084 on Build GRCh38
rs563581711 Chr.12: 3638151-3638151 on Build GRCh38
rs958929900 Chr.12: 3638139-3638139 on Build GRCh38
rs755692821 Chr.12: 3638189-3638189 on Build GRCh38
rs764585207 Chr.12: 3638228-3638233 on Build GRCh38
rs1006431163 Chr.12: 3638198-3638198 on Build GRCh38
rs1054103587 Chr.12: 3638215-3638215 on Build GRCh38
rs1052160691 Chr.12: 3638157-3638157 on Build GRCh38
rs532461381 Chr.12: 3638159-3638159 on Build GRCh38
rs762797657 Chr.12: 3638103-3638103 on Build GRCh38
rs760934861 Chr.12: 3638159-3638159 on Build GRCh38
rs777429692 Chr.12: 3638196-3638196 on Build GRCh38
rs370926137 Chr.12: 3638220-3638220 on Build GRCh38
rs942336076 Chr.12: 3638209-3638209 on Build GRCh38
rs1054176132 Chr.12: 3638049-3638049 on Build GRCh38
rs986366977 Chr.12: 3638056-3638056 on Build GRCh38
rs954715791 Chr.12: 3638053-3638053 on Build GRCh38
rs773065132 Chr.12: 3638108-3638108 on Build GRCh38
rs145536162 Chr.12: 3638118-3638118 on Build GRCh38
rs758755969 Chr.12: 3638207-3638207 on Build GRCh38

View Full Product Details